Skip to Content
Merck
All Photos(1)

Key Documents

17971

Supelco

Arsenic(III) oxide

reference material for titrimetry, certified by BAM, >99.5%

Synonym(s):

Arsenic trioxide, Arsenous acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
As2O3
CAS Number:
Molecular Weight:
197.84
EC Number:
MDL number:
UNSPSC Code:
41116107
eCl@ss:
38080102
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

reference material

Quality Level

Assay

>99.5%

quality

certified by BAM

technique(s)

titration: suitable

application(s)

agriculture
environmental

format

neat

SMILES string

O=[As]O[As]=O

InChI

1S/As2O3/c3-1-5-2-4

InChI key

IKWTVSLWAPBBKU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Arsenic(III) oxide is a volumetric standard produced under rigorous conditions and measured with the highest possible precision. Its content determination and homogeneity assessment are carried out using volumetric redox titration as the primary analysis method.

Application

The reference material is to be used as a reductometric standard for titer determinations in redox titrations.

Features and Benefits

  • Available as a solid in a secure glass bottle to ensure its stability for the entire shelf life until opened.
  • Traceable to NIST standard reference material
  • Offers accurate titer determinations
  • Accompanied by a certificate of analysis (CoA)

Analysis Note

Exact concentration, expiry date and detailed information on certification can be found on the certificate and certification report, included in each package. Content and expiry date on label.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A - Eye Dam. 1 - Skin Corr. 1B - STOT RE 1

Target Organs

Respiratory system,Cardio-vascular system,Gastrointestinal tract

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael Ash, Irene Ash
Handbook of Preservatives, 281 -281 (2004)
Arsenic?glutathione complexes?their stability in solution and during separation by different HPLC modes.
Raab, Andrea, et al.
Journal of Analytical Atomic Spectrometry, 19.1, 183-190 (2004)
Cui Li et al.
Toxicology letters, 219(3), 223-230 (2013-04-02)
Arsenic trioxide (As2O3; ATO) is clinically effective in treating acute promyelocytic leukemia (APL); however, it frequently causes cardiotoxic effects. This study was designed to investigate whether ATO could induce apoptosis of cardiac fibroblasts (CFs) that play very important roles in
Ling-Huei Yih et al.
Toxicology and applied pharmacology, 267(3), 228-237 (2013-01-29)
Accumulated evidence has revealed a tight link between arsenic trioxide (ATO)-induced apoptosis and mitotic arrest in cancer cells. AKT, a serine/threonine kinase frequently over-activated in diverse tumors, plays critical roles in stimulating cell cycle progression, abrogating cell cycle checkpoints, suppressing
Masamitsu Yanada et al.
Blood, 121(16), 3095-3102 (2013-02-16)
The optimal treatments for relapsed acute promyelocytic leukemia (APL) remain equivocal. We conducted a phase 2 study to evaluate the efficacy and feasibility of a sequential treatment consisting of induction and consolidation with arsenic trioxide (ATO), peripheral blood stem cell

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service