Skip to Content
Merck

Skip To

16596

Iron Standard for AAS

TraceCERT®, 1 g/L Fe in nitric acid (nominal concentration)

Synonym(s):

Iron Standard for AAS (1 g/L Fe in 2% nitric acid)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

TraceCERT®

grade

certified reference material, TraceCERT®

concentration

1 g/L Fe in nitric acid (nominal concentration)

technique(s)

AAS: suitable, ICP: suitable

application(s)

industrial qc
pharmaceutical

format

single component solution

General description

This certified reference material (CRM) is produced in accordance with ISO 17034 and characterized in accordance with ISO/IEC 17025. This CRM is traceable to SI unit kg and measured against primary material from a National Metrology Institute (NMI), e.g. NIST.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.

Application

Iron standard has been used as standard stock solution in flame atomic absorption spectrometry (FAAS) method for determining iron and polybdenum in dietetic pharmaceutical preparation.
Iron standard has been used as standard stock solution in flame atomic absorption spectrometry (FAAS) method for determining iron and polybdenum in dietetic pharmaceutical preparation.[1]

Analysis Note

1 g/L Fe in 2% nitric acid, prepared with high purity Fe metal, HNO3 and water

Other Notes

For a complete product listing of our TraceCERT® range of CRMs for ICP and AAS, technical information and example certificates, please visit our ICP & AAS standards website

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
941177424416259
grade

certified reference material, TraceCERT®

grade

certified reference material, TraceCERT®

grade

certified reference material, TraceCERT®

grade

certified reference material, TraceCERT®

technique(s)

AAS: suitable, ICP: suitable

technique(s)

AAS: suitable

technique(s)

AAS: suitable

technique(s)

AAS: suitable

format

single component solution

format

single component solution

format

single component solution

format

single component solution

application(s)

industrial qc
pharmaceutical

application(s)

industrial qc
pharmaceutical

application(s)

industrial qc
pharmaceutical

application(s)

industrial qc
pharmaceutical

product line

TraceCERT®

product line

TraceCERT®

product line

TraceCERT®

product line

TraceCERT®

Quality Level

300

Quality Level

300

Quality Level

300

Quality Level

300


pictograms

Corrosion

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter

Storage Class

12 - Non Combustible Liquids



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



E Canfranc et al.
Journal of pharmaceutical and biomedical analysis, 25(1), 103-108 (2001-03-29)
Methods for the determination of iron and molybdenum in a dietetic pharmaceutical preparation by flame atomic absorption spectrometry (FAAS) after dry ashing at 600 degrees C have been validated. Linearity, precision, accuracy, detection and quantification limits, specificity and robustness have
Paula Barjona do Nascimento Coutinho et al.
Molecular biotechnology, 61(5), 305-316 (2019-01-19)
Microalgae are in the focus for the production of recombinant proteins in research and potential commercial application. Inducible promoters represent important tools that potentially allow the expression of recombinant proteins at higher rates. In general, they are used to separate
Mary E Peek et al.
Interdisciplinary perspectives on infectious diseases, 2012, 843509-843509 (2012-09-14)
Pseudomonas aeruginosa is the most common pathogen that persists in the cystic fibrosis lungs. Bacteria such as P. aeruginosa secrete siderophores (iron-chelating molecules) and the host limits bacterial growth by producing neutrophil-gelatinase-associated lipocalin (NGAL) that specifically scavenges bacterial siderophores, therefore



Global Trade Item Number

SKUGTIN
393851-1G04061831982114
16596-250ML04061837109775

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters