Skip to Content
Merck
All Photos(1)

Key Documents

16033

Sigma-Aldrich

1,2-Propanediol

meets analytical specification of Ph. Eur., BP, USP, ≥99.5%

Synonym(s):

1,2-PDO, Propylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH2OH
CAS Number:
Molecular Weight:
76.09
Beilstein:
1340498
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

vapor density

2.62 (vs air)

Quality Level

vapor pressure

0.08 mmHg ( 20 °C)

Assay

≥99.5%

form

liquid

autoignition temp.

779 °F

quality

meets analytical specification of Ph. Eur., BP, USP

shelf life

24 mo.

expl. lim.

12.5 %

impurities

acidic reac. impurities, complies
organic volatile impurities, complies (GC)
oxidizing substances, complies
reducing matter, complies
residual solvents, complies
≤0.0005% heavy metals (as Pb)
≤0.002% free alkali (as NaOH)
≤0.003% free acid (as CH3COOH)
≤0.2% water (Karl Fischer)

ign. residue

≤0.005% (as SO4)

refractive index

n20/D 1.431-1.433
n20/D 1.432 (lit.)

bp

187 °C (lit.)

mp

−60 °C (lit.)

density

1.036 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

suitability

complies for appearance of solution

functional group

hydroxyl

SMILES string

CC(O)CO

InChI

1S/C3H8O2/c1-3(5)2-4/h3-5H,2H2,1H3

InChI key

DNIAPMSPPWPWGF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,2-Propanediol is an 1,2-alkanediol. It can be obtained from glycerol, via hydrogenolysis in the presence of Ru/C and Amberlyst ion-exchange resins.[1]
1,2-Propanediol is an important petrochemical that can be prepared by the hydrogenolysis of glycerol.[1] It is mainly used as a cryoprotectant for biological samples[2] and also as an additive in food, cosmetic and pharmaceutical industries.[3]

Application

1,2-Propanediol may be employed as a cryoprotective agent for the preservation (by cooling to sub-zero temperatures) of human embryos[4] and and oocytes (human and mouse).[2]

Packaging

bottled under inert gas

Other Notes

The article number 16033-4X5L will be discontinued. Please order the single bottle 16033-5L which is physically identical with the same exact specifications.
The article number 16033-6X1L will be discontinued. Please order the single bottle 16033-1L which is physically identical with the same exact specifications.
The article number 16303-4X2.5L-R will be discontinued. Please order the single bottle 16303-2.5L-R which is physically identical with the same exact specifications.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aqueous phase hydrogenolysis of glycerol to 1, 2-propanediol without external hydrogen addition.
Roy D, et al.
Catalysis Today, 156(1), 31-37 (2010)
B Lassalle et al.
Fertility and sterility, 44(5), 645-651 (1985-11-01)
Eighty-five human embryos fertilized in vitro were frozen and thawed with 1,2 propanediol as a cryoprotective agent. The effects of viability of in vitro culture duration, stage, and morphologic appearance of embryos were examined after thawing. Survival was higher for
Glycerol hydrogenolysis to 1, 2-propanediol catalyzed by a heat-resistant ion-exchange resin combined with Ru/C.
Miyazawa T, et al.
Applied Catalysis A: General, 329, 30-35 (2007)
D A Gook et al.
Human reproduction (Oxford, England), 8(7), 1101-1109 (1993-07-01)
Human and mouse oocytes were cryopreserved by a slow freeze, rapid thaw method, using propanediol (PROH) as the cryoprotectant. A simulated cryopreservation was also included in the study to detect the level of damage attributable to the PROH alone. Comparison
Yusha Imtiaz et al.
Nanomaterials (Basel, Switzerland), 10(8) (2020-08-23)
Polysaccharides have been shown to have immunomodulatory properties. Modulation of the immune system plays a crucial role in physiological processes as well as in the treatment and/or prevention of autoimmune and infectious diseases. Cellulose nanocrystals (CNCs) are derived from cellulose

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service