Skip to Content
Merck
All Photos(1)

Key Documents

15889

Supelco

Density Standard 1623 kg/m3

H&D Fitzgerald Ltd. Quality

Synonym(s):

Tetrachloroethylene, PCE, Perchloroethylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CCl2=CCl2
CAS Number:
Molecular Weight:
165.83
Beilstein:
1361721
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

H&D Fitzgerald Ltd. Quality
analytical standard

vapor density

5.83 (vs air)

vapor pressure

13 mmHg ( 20 °C)
19 mmHg ( 25 °C)

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

H&D Fitzgerald Ltd.

refractive index

n20/D 1.505 (lit.)

bp

121 °C (lit.)

mp

−22 °C (lit.)

density

1.623 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
industrial qc
personal care
petroleum
pharmaceutical (small molecule)

format

mixture

SMILES string

Cl\C(Cl)=C(\Cl)Cl

InChI

1S/C2Cl4/c3-1(4)2(5)6

InChI key

CYTYCFOTNPOANT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

This density CRM is used for the calibration of all types of laboratory density meters in the test temperature range of 10 to 50 °C.

Features and Benefits

  • Manufactured and verified in an ISO 17025 accredited laboratory, as per UKAS
  • Carries an associated uncertainty of +/- 0.010 Kg/m3
  • Provided in a sealed glass ampoule of 10 mL to assure safe delivery
  • Accompanied with a certificate of analysis (CoA) having information on the expiry date for an effective audit

Signal Word

Warning

Hazard Classifications

Aquatic Chronic 2 - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thermophysical Properties of 1-Butyl-1-methyl-pyrrolidinium Dicyanamide+ H2O Mixtures.
Cumicheo, M. C., et al.
Journal of Chemical and Engineering Data, 60.12, 3766-3775 (2015)
B Z Fathepure et al.
Applied and environmental microbiology, 54(12), 2976-2980 (1988-12-01)
Tetrachloroethylene (perchloroethylene, PCE) is a suspected carcinogen and a common groundwater contaminant. Although PCE is highly resistant to aerobic biodegradation, it is subject to reductive dechlorination reactions in a variety of anaerobic habitats. The data presented here clearly establish that
T M Vogel et al.
Applied and environmental microbiology, 49(5), 1080-1083 (1985-05-01)
Tetrachloroethylene (PCE) and trichloroethylene (TCE), common industrial solvents, are among the most frequent contaminants found in groundwater supplies. Due to the potential toxicity and carcinogenicity of chlorinated ethylenes, knowledge about their transformation potential is important in evaluating their environmental fate.
Julie C Chambon et al.
Biotechnology and bioengineering, 110(1), 1-23 (2012-08-29)
Reductive dechlorination is a major degradation pathway of chlorinated ethenes in anaerobic subsurface environments, and reactive kinetic models describing the degradation process are needed in fate and transport models of these contaminants. However, reductive dechlorination is a complex biological process
Krista Yorita Christensen et al.
Journal of occupational and environmental medicine, 55(2), 198-208 (2012-11-14)
To evaluate the association between exposure to chlorinated solvents and cancer. We conducted a case-control study of occupational exposures and cancer in Montreal, Quebec, Canada, including 3730 cancer cases and 533 population controls. Occupational exposures were derived using a combination

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service