11133
Antimony(V) sulfide
technical, ≥60% Sb basis
Synonym(s):
Antimony pentasulfide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
Sb2S5
CAS Number:
Molecular Weight:
403.85
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
grade
technical
form
solid
reaction suitability
core: antimony
reagent type: catalyst
concentration
≥60% Sb
SMILES string
S=[Sb](=S)S[Sb](=S)=S
InChI
1S/5S.2Sb
InChI key
PPKVREKQVQREQD-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Flam. Sol. 1
Storage Class Code
4.1B - Flammable solid hazardous materials
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Brigitta Müller et al.
Planta, 249(3), 751-763 (2018-11-02)
Fe uptake machinery of chloroplasts prefers to utilise Fe(III)-citrate over Fe-nicotianamine complexes. Iron uptake in chloroplasts is a process of prime importance. Although a few members of their iron transport machinery were identified, the substrate preference of the system is
[Substantiation of maximum permissible levels of antimony trioxide and pentasulide in the atmospheric air of inhabitated places].
N V Grin' et al.
Gigiena i sanitariia, (4)(4), 68-69 (1989-04-01)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service