Skip to Content
Merck
All Photos(1)

Key Documents

09668

Sigma-Aldrich

N,N,N′,N′-Tetramethyl-O-(N-succinimidyl)uronium hexafluorophosphate

≥99.0% (TLC/N)

Synonym(s):

O-(N-Succinimidyl)-N,N,N′,N′-tetramethyluronium hexafluorophosphate, HSTU

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H16F6N3O3P
CAS Number:
Molecular Weight:
359.21
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0% (TLC/N)

form

crystals

reaction suitability

reaction type: Coupling Reactions

mp

218-221 °C (dec.)

solubility

acetonitrile: 0.5 g/mL, slightly hazy, colorless

application(s)

peptide synthesis

storage temp.

−20°C

SMILES string

F[P-](F)(F)(F)(F)F.CN(C)C(\ON1C(=O)CCC1=O)=[N+](\C)C

InChI

1S/C9H16N3O3.F6P/c1-10(2)9(11(3)4)15-12-7(13)5-6-8(12)14;1-7(2,3,4,5)6/h5-6H2,1-4H3;/q+1;-1

InChI key

STWZCCVNXFLDDD-UHFFFAOYSA-N

General description

N,N,N′,N′-Tetramethyl-O-(N-succinimidyl)uronium hexafluorophosphate can be used to prepare N-succinimidyl 4-[18F]fluorobenzoate ([18F]-SFB), a prosthetic group for the fluorination of biomolecules for PET imaging.

Application

Reactant for:
Synthesis of liposomal contrast agents for magnetic resonance imaging
Synthesis of thiol-reactive Cy5 derivatives
Synthesis of protein labeling molecules

Other Notes

Coupling reagent for peptide synthesis and the formation of other amides. Even in aqueous solution yields are excellent

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

18F-nanobody for PET imaging of HER2 overexpressing tumors
Xavier C, et al.
Nuclear Medicine and Biology, 43, 247-252 (2016)
Synthesis of 18F-labeled membrane-penetrating peptide as PET tracer
Tang G and Wang X
Journal of Nuclear Medicine, 49, 97P-97P (2008)
PET imaging of macrophage mannose receptor-expressing macrophages in tumor stroma using 18F-radiolabeled camelid single-domain antibody fragments
Blykers A, et al.
Journal of Nuclear Medicine, 56, 1265-1271 (2015)
Synthesis and in vivo evaluation of p-18F-Fluorohippurate as a new radiopharmaceutical for assessment of renal function by PET
Awasthi V, et al.
Journal of Nuclear Medicine, 52, 147-153 (2011)
Virginia Wycisk et al.
ChemistryOpen, 6(3), 437-446 (2017-06-24)
Herein, we present a new synthetic route to cyanine-based heterobifunctional dyes and their application as fluorescent linkers between polymers and biomolecules. The synthesized compounds, designed in the visible spectral range, are equipped with two different reactive groups for highly selective

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service