Skip to Content
Merck

Skip To

08069

modified Tryptic Soy Broth- Dehydrated Culture Media

NutriSelect® Plus, with bile salts mixture, according to ISO 16654:2001, powder, selective for enterovirulent E. coli (EEC) O157:H7

Synonym(s):

Modified Soyabean Bile Broth, Modified soybean bile broth base, Modified tryptone soy broth, Tryptic soy broth, modified, mTSB

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.85
UNSPSC Code:
41171607
Manufacturer/tradename:
NutriSelect® Plus
Agency:
according to ISO 16654:2001
Form:
powder
Suitability:
selective for enterovirulent E. coli (EEC) O157:H7
Application(s):
food and beverages
microbiology
pathogen testing
vitamins, nutraceuticals, and natural products
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


agency

according to ISO 16654:2001

Quality Segment

sterility

non-sterile

form

powder

shelf life

limited shelf life, expiry date on the label

composition

casein enzymic hydrolysate, 17 g/L , dipotassium hydrogen phosphate, 4 g/L , D-glucose, 2.5 g/L , papaic digest of soyabean meal, 3 g/L , sodium chloride, 5 g/L , with bile salts mixture

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

7.3±0.2 (25 °C)

application(s)

food and beverages
microbiology
pathogen testing
vitamins, nutraceuticals, and natural products

suitability

selective for enterovirulent E. coli (EEC) O157:H7

Application

Modified tryptone soy broth is recommended as an enrichment medium for the detection of Escherichia coli O157:H7 from food and animal feeding stuffs.

Preparation Note

Suspend 33 grams of Modified tryptone soy broth in 1000 ml distilled water. Heat if necessary to dissolve the medium completely. Sterilize by autoclaving at 15 lbs pressure (121 °C) for 15 minutes. Cool to 45-50 °C and aseptically add rehydrated contents of 1 vial of VCC Selective Supplement (Cat. No. No. 80704). Mix well and dispense as desired.

Other Notes

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.400111.092051.46432
application(s)

food and beverages
microbiology
pathogen testing
vitamins, nutraceuticals, and natural products

application(s)

food and beverages, microbiology
pathogen testing

application(s)

food and beverages
microbiology
pathogen testing

application(s)

cosmetics
food and beverages
pharmaceutical
sterility testing

technique(s)

microbiological culture: suitable

technique(s)

-

technique(s)

microbiological culture: suitable

technique(s)

-

manufacturer/tradename

NutriSelect® Plus

manufacturer/tradename

GranuCult® prime

manufacturer/tradename

GranuCult® prime

manufacturer/tradename

-

agency

according to ISO 16654:2001

agency

according to USDA-FSIS

agency

FDA-BAM, ISO 16654

agency

EP 2.6.1, JP 4.06, USP 71

form

powder

form

granular

form

granular

form

liquid

suitability

selective for enterovirulent E. coli (EEC) O157:H7

suitability

Escherichia coli O157:H7, Escherichia coli (non-O157 Shiga Toxin-Producing (STEC)), Salmonella spp.

suitability

Escherichia coli (O157:H7 ), selective for Escherichia coli (EHEC), urea-degrading bacteria

suitability

nonselective for


Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters