Skip to Content
Merck
All Photos(2)

Key Documents

06595

Supelco

5-Aminoimidazole-4-carboxamide-1-β-D-ribofuranosyl 5′-monophosphate

analytical standard

Synonym(s):

AICAR monophosphate, N1-(β-D-5′-Phosphoribofuranosyl)-5-aminoimidazole-4-carboxamide, NSC 283955, NSC 292227, ZMP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H15N4O8P
CAS Number:
Molecular Weight:
338.21
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

optical activity

[α]/D -69.0±3.0°, c = 0.1 in sodium bicarbonate

shelf life

limited shelf life, expiry date on the label

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

O[C@H]1[C@@H](O)[C@H](N2C=NC(C(N)=O)=C2N)O[C@@H]1COP(O)(O)=O

InChI

1S/C9H15N4O8P/c10-7-4(8(11)16)12-2-13(7)9-6(15)5(14)3(21-9)1-20-22(17,18)19/h2-3,5-6,9,14-15H,1,10H2,(H2,11,16)(H2,17,18,19)/t3-,5-,6-,9-/m1/s1

InChI key

NOTGFIUVDGNKRI-UUOKFMHZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

5-Aminoimidazole-4-carboxamide-1-β-D-ribofuranosyl 5′-monophosphate (AICAR monophosphate) is an intermediate metabolite in the purine de novo synthesis pathway and a potent activator of the human AMP-activated protein kinase (AMPK) activity in vitro. It can play an important regulatory role.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joshua D Stone et al.
American journal of physiology. Heart and circulatory physiology, 304(3), H369-H381 (2012-12-04)
Vascular smooth muscle cell (VSMC) activation promotes a synthetic phenotype that underlies many vessel growth disorders. In this regard it has been suggested that the metabolic sensor adenosine 5'-monophosphate-activated protein kinase (AMPK) has significant antigrowth and antimetastatic properties and may
Hans P M M Lauritzen et al.
Diabetes, 62(9), 3081-3092 (2013-06-14)
Recent studies suggest that interleukin 6 (IL-6) is released from contracting skeletal muscles; however, the cellular origin, secretion kinetics, and signaling mechanisms regulating IL-6 secretion are unknown. To address these questions, we developed imaging methodology to study IL-6 in fixed
Licht Miyamoto et al.
American journal of physiology. Cell physiology, 305(12), C1214-C1222 (2013-10-04)
Physical exercise has potent therapeutic and preventive effects against metabolic disorders. A number of studies have suggested that 5'-AMP-activated protein kinase (AMPK) plays a pivotal role in regulating carbohydrate and lipid metabolism in contracting skeletal muscles, while several genetically manipulated
Janine Dokas et al.
Endocrinology, 154(10), 3502-3514 (2013-07-31)
In the obesity-resistant SJL mouse strain, we previously identified a naturally occurring loss-of-function mutation in the gene for Tbc1d1. Characterization of recombinant inbred mice that carried the Tbc1d1(SJL) allele on a C57BL/6J background indicated that loss of TBC1D1 protects from
5-Aminoimidazole-4-carboxamide-1-beta-D-ribofuranosyl 5'-monophosphate (AICAR), a highly conserved purine intermediate with multiple effects
Daignan-Fornier B and Pinson B
Metabolites, 2(2), 292-302 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service