Skip to Content
Merck
All Photos(1)

Documents

06233

Sigma-Aldrich

Aluminum phosphate monobasic

purum, ≥95.0% (gravimetric)

Synonym(s):

Aluminum dihydrogen orthophosphate, Aluminum dihydrogen phosphate, Aluminum hydrogen phosphate, prim-Aluminum phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Al(H2PO4)3
CAS Number:
Molecular Weight:
317.94
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Quality Level

Assay

≥95.0% (gravimetric)

form

powder

reaction suitability

core: aluminum
reagent type: catalyst

loss

≤20% loss on ignition, 800 °C

SMILES string

[Al+3].OP(O)([O-])=O.OP(O)([O-])=O.OP(O)([O-])=O

InChI

1S/Al.3H3O4P/c;3*1-5(2,3)4/h;3*(H3,1,2,3,4)/q+3;;;/p-3

InChI key

RGPUVZXXZFNFBF-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Aluminum phosphate monobasic can be used as a combining material of refractories. It can be used as a precursor for the synthesis of:
  • Microwave absorbing silicon carbide (SiC) fiber-reinforced SiC matrix composites (SiCf/SiC).
  • Aluminum doped hydroxyapatite crystals.
  • Phosphate insulating coating to improve the corrosion resistance of silicon steel.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Improvement dielectric and microwave properties of SiCf/SiC-AlPO4 composites prepared by precursor infiltration and pyrolysis process.
Hu Y, et al.
Journal of alloys and compounds, 699(12), 498-504 (2017)
Effects of ammonium tungstate on the properties of insulating coating for grain-oriented silicon steel.
Wang C, et al.
Materials Science and Engineering, B, 228(31), 109-116 (2018)
The crystal structure and chemical state of aluminum-doped hydroxyapatite by experimental and first principles calculation studies.
Wang M, et al.
Physical Chemistry Chemical Physics, 18(31), 21789-21796 (2016)
Reaction of monoaluminum phosphate with alumina and magnesia.
Ando J, et al.
J. Ceram. Soc. Jpn., 82(12), 644-649 (1974)
Belma Ljutic et al.
Vaccine, 30(19), 2981-2988 (2012-03-03)
We investigated the immunogenicity, stability and adsorption properties of an experimental pneumococcal vaccine composed of three protein vaccine antigens; Pneumococcal histidine triad protein D, (PhtD), Pneumococcal choline-binding protein A (PcpA) and genetically detoxified pneumolysin D1 (PlyD1) formulated with aluminum salt

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service