Skip to Content
Merck
All Photos(1)

Key Documents

04704

Supelco

Glycidamide

analytical standard

Synonym(s):

2,3-Epoxypropanamide, Oxiranecarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H5NO2
CAS Number:
Molecular Weight:
87.08
Beilstein:
227 163 2
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95% (qNMR)

shelf life

limited shelf life, expiry date on the label

application(s)

environmental

format

neat

storage temp.

2-8°C

SMILES string

NC(=O)C1CO1

InChI

1S/C3H5NO2/c4-3(5)2-1-6-2/h2H,1H2,(H2,4,5)

InChI key

FMAZQSYXRGRESX-UHFFFAOYSA-N

Related Categories

General description

Glycidamide, an active epoxide metabolite of acrylamide with potential biological activity, can be generated in vivo. Its mutagenic and genotoxic nature is well-known.

Application

Glycidamide may be used as an analytical standard for the determination of the analyte in human and animal biological samples by various chromatography techniques.

Other Notes

In the summer of 2008 research scientists at the Technical University of Munich besides acrylamide detected glycidamide in potato crisps. Glycidamide is a metabolite of acrylamide.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Malene Outzen et al.
The British journal of nutrition, 105(9), 1381-1387 (2011-01-29)
Acrylamide (AA) is a probable human carcinogen that is formed in heat-treated carbohydrate-rich foods. The validity of FFQ to assess AA exposure has been questioned. The aim of the present cross-sectional study was to investigate dietary determinants of Hb-AA and
Anna C Vikström et al.
Toxicological sciences : an official journal of the Society of Toxicology, 119(1), 41-49 (2010-10-19)
For assessment of cancer risk from acrylamide (AA) exposure through food, the relation between intake from food in humans and the in vivo doses (area under the concentration-time curve, AUC) of AA (AUC-AA) and of its genotoxic metabolite glycidamide (GA)
Tae Hwan Kim et al.
Talanta, 131, 46-54 (2014-10-05)
Acrylamide (AA) is a heat-generated food toxicant formed when starchy foods are fried or baked. This study describes a simple and sensitive liquid chromatography-tandem mass spectrometry assay for the simultaneous quantification of AA and its active metabolite, glycidamide (GA) in
Simultaneous determination of acrylamide, its metabolite glycidamide and antipyrine in human placental perfusion fluid and placental tissue by liquid chromatography-electrospray tandem mass spectrometry
Annola K, et al.
Journal of Chromatography. B, Biomedical Applications, 876(2), 191-197 (2008)
Hans von Stedingk et al.
Chemical research in toxicology, 24(11), 1957-1965 (2011-09-03)
The knowledge about fetal exposure to acrylamide/glycidamide from the maternal exposure through food is limited. Acrylamide, glycidamide, and ethylene oxide are electrophiles and form adducts with hemoglobin (Hb), which could be used for in vivo dose measurement. In this study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service