Skip to Content
Merck

Skip To

04071

Alkane standard solution C21-C40

analytical standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41116107
EC Number:
203-625-9
NACRES:
NA.24
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.497

application(s)

petroleum

format

multi-component solution

Application

Alkane standard solution C21-C40 has been used as an analytical standard, to evaluate the limits of detection and relative sensitivities involved in the functionalization of several saturated hydrocarbons using nitrogen ion insertion reactions in mass spectrometry (MS).

Other Notes

contains C21, C22, C23, C24, C25, C26, C27, C28, C29, C30, C31, C32, C33, C34, C35, C36, C37, C38, C39, C40, 40 mg/l each, in toluene

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
0407040147-U49452-U
application(s)

petroleum

application(s)

petroleum

application(s)

petroleum

application(s)

petroleum

format

multi-component solution

format

multi-component solution

format

multi-component solution

format

multi-component solution

grade

analytical standard

grade

analytical standard

grade

-

grade

certified reference material

refractive index

n20/D 1.497

refractive index

n20/D 1.375

refractive index

-

refractive index

-

Quality Level

100

Quality Level

100

Quality Level

300

Quality Level

300

shelf life

limited shelf life, expiry date on the label

shelf life

-

shelf life

-

shelf life

-


signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 3

flash_point_f

39.9 °F - closed cup

flash_point_c

4.4 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters