Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C26H33NO4
CAS Number:
Molecular Weight:
423.54
UNSPSC Code:
12352209
NACRES:
NA.26
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
4887890
Quality Segment
assay
≥98.0% (HPLC)
form
powder
reaction suitability
reaction type: Fmoc solid-phase peptide synthesis
color
white
application(s)
peptide synthesis
functional group
Fmoc
storage temp.
2-8°C
SMILES string
OC(=O)CCCCCCCCCCNC(=O)OCC1c2ccccc2-c3ccccc13
InChI
1S/C26H33NO4/c28-25(29)17-7-5-3-1-2-4-6-12-18-27-26(30)31-19-24-22-15-10-8-13-20(22)21-14-9-11-16-23(21)24/h8-11,13-16,24H,1-7,12,17-19H2,(H,27,30)(H,28,29)
InChI key
IMEVDILDSCXKJW-UHFFFAOYSA-N
Other Notes
Alkyl spacer employed in peptide synthesis.
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
