Skip to Content
Merck
All Photos(1)

Key Documents

03835

Supelco

γ-Aminobutyric acid

analytical standard

Synonym(s):

3-Carboxypropylamine, 4-Aminobutanoic acid, GABA, Piperidic acid, Piperidinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2(CH2)3COOH
CAS Number:
Molecular Weight:
103.12
Beilstein:
906818
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

analyte chemical class(es)

amino acids, peptides, proteins

technique(s)

HPLC: suitable

impurities

≤1.0% water

mp

195 °C (dec.) (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

NCCCC(O)=O

InChI

1S/C4H9NO2/c5-3-1-2-4(6)7/h1-3,5H2,(H,6,7)

InChI key

BTCSSZJGUNDROE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

γ-Aminobutyric acid is a chief inhibitory neurotransmitter, which is found in the cerebellum, hypothalamus, thalamus and hippocampus. It is formed via the decarboxylation of L-glutamate catalyzed by the enzyme, glutamic acid decarboxylase(GAD).

Biochem/physiol Actions

Major inhibitory neurotransmitter in brain; GABAA and GABAB receptor agonist; increases Cl conductance.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wen-Zhao Cui et al.
Biology, 10(9) (2021-09-29)
Diapause is a developmental transition in insects based on seasonal adaptation to adversity; it is regulated by a circadian clock system and the endocrine system. However, the molecular node and its mechanism underlying the effects of these systems are still
Medicinal Chemistry: A Molecular and Biochemical Approach (2005)
Antonela G Garzón et al.
Journal of food science and technology, 55(8), 3188-3198 (2018-08-02)
Two cultivars of sorghum were germinated at 25 or 30 °C for 1, 2, or 3 days to investigate the evolution of γ-Aminobutyric acid (GABA), total free phenolic compounds (FPC), hydroxycinnamic acid derivatives, free amino acid (FAA) profile, and antioxidant activity during
Jaisan Islam et al.
International journal of molecular sciences, 22(16) (2021-08-28)
The nucleus accumbens core (NAcc) is an important component of brain reward circuitry, but studies have revealed its involvement in pain circuitry also. However, its effect on trigeminal neuralgia (TN) and the mechanism underlying it are yet to be fully
Shang-Feng Gao et al.
The Neuroscientist : a review journal bringing neurobiology, neurology and psychiatry, 17(1), 124-144 (2010-03-20)
Stress response and depression have a significant impact on modern society. Although the symptoms are well characterized, the molecular mechanisms underlying depression are largely unknown. The monoamine hypothesis, which postulates dysfunctional noradrenergic and serotonergic systems as the underlying primary cause

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service