Skip to Content
Merck
All Photos(1)

Key Documents

02799

Supelco

Tetramethylammonium sulfate

suitable for ion pair chromatography, LiChropur, ≥99.0% (T)

Synonym(s):

Bis(tetramethylammonium) sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)4N]2(SO4)
CAS Number:
Molecular Weight:
244.35
Beilstein:
3918039
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NB.21

description

cationic

Quality Level

Assay

≥99.0% (T)

form

powder

quality

LiChropur

technique(s)

ion pair chromatography: suitable

λ

10 % in H2O

UV absorption

λ: 210 nm Amax: ≤0.1
λ: 220 nm Amax: ≤0.04
λ: 230 nm Amax: ≤0.03
λ: 260 nm Amax: ≤0.02
λ: 500 nm Amax: ≤0.02

suitability

corresponds to standard for RP gradient test
corresponds to standard for filter test

SMILES string

C[N+](C)(C)C.C[N+](C)(C)C.[O-]S([O-])(=O)=O

InChI

1S/2C4H12N.H2O4S/c3*1-5(2,3)4/h2*1-4H3;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

KJFVITRRNTVAPC-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application


  • Separation of Gd-humic complexes and Gd-based magnetic resonance imaging contrast agent in river water with QAE-Sephadex A-25 for the fractionation analysis.: This study highlights the application of Tetramethylammonium sulfate in the separation of gadolinium complexes from river water using QAE-Sephadex A-25. The research demonstrates the effectiveness of this approach in environmental analytical chemistry, providing insights into the behavior and fate of contrast agents in natural water systems (Matsumiya et al., 2014).

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yasuyuki Ishida et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 27(10), 1053-1056 (2011-10-12)
Thermochemolysis-gas chromatography in the presence of tetramethylammonium acetate was applied to the direct determination of terephthalic acid (TPA) contained in solid decomposition products obtained from the hydrothermal recycling process of poly(ethylene terephthalate) (PET). On the chromatograms of the hydrothermal decomposition
Qin Chen et al.
Bioresource technology, 117, 186-192 (2012-05-23)
The aim of this study was to explore the synergies of laccase, a ligninolytic enzyme, with cellulose and hemicellulase amendments on ensiled corn stover. Molecular signals of lignin decomposition were observed by tetramethylammonium hydroxide thermochemolysis and gas chromatography-mass spectroscopy (TMAH-GC-MS)
Hui Chong et al.
Langmuir : the ACS journal of surfaces and colloids, 28(4), 2091-2098 (2011-11-08)
A new water-soluble conjugated polymer containing fluorene and boron-dipyrromethene repeat units in the backbones (PBF) that exhibits red emission was synthesized and characterized. Cationic PBF forms uniform nanoparticles with negatively charged disodium salt 3,3'-dithiodipropionic acid (SDPA) in aqueous solution through
Federica Maraschi et al.
Analytical biochemistry, 426(2), 142-146 (2012-05-01)
This article presents the first results demonstrating that total silicon trace concentration in human ventricular whole blood may be used as a further marker in the diagnosis of drowning. The difference in silicon content between the left and right ventricles
Man Xu et al.
Environmental science & technology, 47(1), 110-118 (2012-06-12)
Elucidating dissolution kinetics and mechanisms at carbonate mineral-water interfaces is essential to many environmental and geochemical processes, including geologic CO(2) sequestration in deep aquifers. In the present work, effects of background electrolytes on dolomite (CaMg(CO(3))(2)) reactivity were investigated by measuring

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service