Skip to Content
Merck
All Photos(1)

Documents

02736

Supelco

2-Methylimidazole

≥95.0% (HPLC), pharmaceutical impurity standard

Synonym(s):

Ondansetron impurity F (PhEur), 2-Methylglyoxaline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6N2
CAS Number:
Molecular Weight:
82.10
Beilstein:
1368
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

vapor pressure

<1 mmHg ( 0 °C)

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

impurities

≤10.0% related substances
≤5.0% solvents
≤5.0% water

bp

267-268 °C (lit.)

mp

142-143 °C (lit.)

application(s)

pharmaceutical (small molecule)

storage temp.

2-8°C

SMILES string

Cc1ncc[nH]1

InChI

1S/C4H6N2/c1-4-5-2-3-6-4/h2-3H,1H3,(H,5,6)

InChI key

LXBGSDVWAMZHDD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Repr. 1B - Skin Corr. 1C

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Janosch Cravillon et al.
Angewandte Chemie (International ed. in English), 50(35), 8067-8071 (2011-07-13)
Prenucleation clusters: in situ synchrotron X-ray scattering with a one-second time resolution revealed the occurrence of nano-sized clusters during the nucleation and early growth of nanocrystals of a zeolitic imidazolate framework (ZIF). The complex crystallization process exhibits similarities with crystallization processes
Jiafeng Geng et al.
Journal of the American Chemical Society, 134(29), 12209-12218 (2012-06-30)
Tryptophan 2,3-dioxygenase (TDO) is a heme-dependent enzyme that catalyzes the oxidative degradation of L-tryptophan (L-Trp) to N-formylkynurenine (NFK). A highly conserved histidine residue in the distal heme pocket has attracted great attention in the mechanistic studies of TDO. However, a
Wenxiang Cao et al.
Biochemistry, 43(22), 7017-7027 (2004-06-02)
We use laser flash photolysis and time-resolved Raman spectroscopy of CO-bound heme complexes to study proximal and distal influences on ligand rebinding kinetics. We report kinetics of CO rebinding to microperoxidase (MP) and 2-methylimidazole ligated Fe protoporphyrin IX in the
Cristina Maccallini et al.
Bioorganic & medicinal chemistry letters, 20(22), 6495-6499 (2010-10-12)
A series of N-substituted acetamidines and 2-methylimidazole derivatives structurally related to W1400 were synthesized and evaluated as Nitric Oxide Synthase (NOS) inhibitors. Analogs with sterically hindering isopropyl and phenyl substituents on the benzylic carbon connecting the aromatic core of W1400
M van Gastel et al.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry, 4(3), 257-265 (1999-08-10)
Frozen solutions of the azurin mutant His117Gly in the presence of excess of methyl-substituted imidazoles have been investigated by electron spin-echo envelope modulation (ESEEM) spectroscopy at 9 GHz. The addition of imidazole is known to reconstitute a blue-copper site and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service