Skip to Content
Merck

02489

2-Chloropropane

analytical standard

Synonym(s):

Isopropyl chloride

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Linear Formula:
(CH3)2CHCl
CAS Number:
Molecular Weight:
78.54
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
EC Number:
200-858-8
Beilstein/REAXYS Number:
1730782
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

grade

analytical standard

Quality Level

vapor pressure

28.11 psi ( 55 °C), 8.59 psi ( 20 °C)

assay

≥99.7% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

refractive index

n20/D 1.378

bp

34-36 °C (lit.)

mp

−118 °C (lit.)

density

0.859 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

storage temp.

2-8°C

SMILES string

CC(C)Cl

InChI

1S/C3H7Cl/c1-3(2)4/h3H,1-2H3

InChI key

ULYZAYCEDJDHCC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

-32.8 °F

flash_point_c

-36 °C

ppe

Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Global Trade Item Number

SKUGTIN
F6605-25G04061832493374
F6605-5G04061832493381
02489-5ML04061838617583

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service