Skip to Content
Merck

01916

Agar

powder, suitable for microbiology

Synonym(s):

Agar-agar, Gum agar

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(C12H18O9)n
CAS Number:
NACRES:
NA.74
UNSPSC Code:
41106212
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

Quality Segment

shelf life

limited shelf life, expiry date on the label

loss

≤10% loss on drying

pH

6-7.5 (1.5% in gel, after autoclaving )

mp

85-89 °C

transition temp

gel point 34-36 °C

gel strength

≥800 g/cm2Nikkan, after autoclaving

anion traces

sulfate (SO42-): ≤2.75%

cation traces

Ca: ≤500 ppm, Mg: ≤200 ppm

application(s)

microbiology

storage temp.

15-25°C

InChI

1S/C14H24O9/c1-5-8(16)13-11(7(21-5)4-20-13)23-14-10(18)12(19-2)9(17)6(3-15)22-14/h5-18H,3-4H2,1-2H3/t5?,6-,7?,8-,9+,10-,11?,12+,13+,14?/m1/s1

InChI key

GYYDPBCUIJTIBM-DYOGSRDZSA-N

General description

Agar is a gelatinous substance, commonly used as a solidifying agent in microbiological media. It provides a stable platform for the growth of microorganisms. This agar is a mixture of Gelidium agar and a small amount of Gracilaria agar. It gives a very clear gel and shows the same good characteristics as the typical pure Gelidium agar.

Analysis Note

Toxic substances: absence
Nephelosi/after autoclaving NTU <8


Still not finding the right product?

Explore all of our products under Agar


Storage Class

11 - Combustible Solids

wgk

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library