Skip to Content
Merck

Skip To

TRIS-RO

Tris base

99.9% (titration), solid, abs., 100 mg/mL, 300 nm ≤0.015

Synonym(s):

Trizma® base, amino-2-(hydroxymethyl)-1,3-propanediol, 2-, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Tris(hydroxymethyl)aminomethane, Trometamol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
NH2C(CH2OH)3
CAS Number:
Molecular Weight:
121.14
UNSPSC Code:
12161705
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
741883
NACRES:
NA.54
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Tris base, =99.9% (titration), abs., 100 mg/mL, 300 nm ≤0.015

description

aminopeptidase substrate

assay

99.9% (titration)

packaging

pkg of 1 kg (10708976001), pkg of 5 kg (11814273001)

manufacturer/tradename

Roche

useful pH range

7-9

pKa (25 °C)

8.1

bp

219-220 °C/10 mmHg (lit.)

mp

167-172 °C (lit.)

absorption

≤0.015 at 300 nm at 100 mg/mL

storage temp.

20-25°C

SMILES string

NC(CO)(CO)CO

InChI

1S/C4H11NO3/c5-4(1-6,2-7)3-8/h6-8H,1-3,5H2

InChI key

LENZDBCJOHFCAS-UHFFFAOYSA-N

Analysis Note

Contaminants: <0.001% PK and aldolase, each, <0.01% GOT, GPT, MDH and myokinase, each

Other Notes

For life science research only. Not for use in diagnostic procedures.
The pH values of all buffers are temperature- and concentration-dependent. For Tris buffers, pH increases about 0.03 unit per °C decrease in temperature, and decreases 0.03-0.05 unit per ten-fold dilution.
For precise applications, use a carefully calibrated pH meter with a glass/calomel combination electrode.

Legal Information

Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
T6791T46619210-OP
assay

99.9% (titration)

assay

≥99.9% (titration)

assay

≥99.9% (titration)

assay

-

storage temp.

20-25°C

storage temp.

-

storage temp.

-

storage temp.

room temp

mp

167-172 °C (lit.)

mp

167-172 °C (lit.)

mp

167-172 °C (lit.)

mp

167-172 °C (lit.)

packaging

pkg of 1 kg (10708976001), pkg of 5 kg (11814273001)

packaging

-

packaging

-

packaging

-

useful pH range

7-9

useful pH range

7-9

useful pH range

7-9

useful pH range

7-9

manufacturer/tradename

Roche

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

Calbiochem®


Still not finding the right product?

Explore all of our products under Tris base


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
1181427300104061838705631
1070897600104061838251794