Skip to Content
Merck

Skip To

NGLYF-RO

Roche

N-Glycosidase F

PNGase F of Flavobacterium meningosepticum, recombinant from E. coli

Synonym(s):

glycosidase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352204
EC Number:
NACRES:
NA.54
Specific activity:
~25000 units/mg protein
Assay:
≥90% (SDS-PAGE)
Recombinant:
expressed in E. coli
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


recombinant

expressed in E. coli

conjugate

(N-linked)

assay

≥90% (SDS-PAGE)

form

solution

specific activity

~25000 units/mg protein

packaging

pkg of 0.1 mL (11365169001 [100 U]), pkg of 0.25 mL (11365177001 [250 U])

manufacturer/tradename

Roche

optimum pH

7.0-8.0

storage temp.

−20°C

General description

Peptide-N-glycosidase F, Peptide-N4-(acetyl-β-glucosaminyl)-asparagine amidase

Application

Use N-glycosidase F to cleave all types of asparagine-bound N-glycans, provided that the amino group as well as the carboxyl group are present in a peptide linkage, and that the oligosaccharide has the minimum length of the chitobiose core unit. The reaction products are ammonia, aspartic acid (in the peptide chain), and the complete oligosaccharide.
Note: N-Glycosidase F, recombinant is also available as a lyophilizate without glycerol.

Physical form

Solution in 50 mM sodium phosphate, 12.5 mM EDTA, 50% glycerol (v/v), pH 7.2

Other Notes

For life science research only. Not for use in diagnostic procedures.
One unit is the enzyme activity which hydrolyzes 1 nmol dabsyl fibrin glycopeptide or 0.2 nmol dansyl fetuin glycopeptide within 1 minute at 37 °C at pH 7.8.

Legal Information

The sale of the Product does not exhaust or grant any rights in third party patents including patents of companies of the F. Hoffmann - La Roche AG group of companies, in particular, for the use of modified antibodies obtained by using the product.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P9120NGLYFLROP7367
description

PNGase F of Flavobacterium meningosepticum, recombinant from E. coli

description

recombinant, expressed in E. coli, set of 100 units nanomolar unit

description

recombinant form of the gene from Flavobacterium meningosepticum

description

BioReagent, ≥95% (SDS-PAGE)

assay

≥90% (SDS-PAGE)

assay

-

assay

≥90% (SDS-PAGE)

assay

≥95% (SDS-PAGE)

specific activity

~25000 units/mg protein

specific activity

≥10 units/mg protein

specific activity

>25000 units/mg protein

specific activity

-

recombinant

expressed in E. coli

recombinant

expressed in E. coli

recombinant

expressed in E. coli

recombinant

-

form

solution

form

-

form

lyophilized

form

powder

optimum pH

7.0-8.0

optimum pH

-

optimum pH

7.0-8.0

optimum pH

~8.6

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C


Still not finding the right product?

Explore all of our products under N-Glycosidase F


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

No data available

flash_point_c

No data available



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library