Skip to Content
Merck

IPTG-RO

Roche

Isopropyl-β-D-thiogalactoside

mol wt 238.3, pkg of 1 g (10724815001), pkg of 5 g (11411446001)

Synonym(s):

Isopropyl β-D-1-thiogalactopyranoside, iptg, IPTG, Isopropyl β-D-thiogalactoside

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Empirical Formula (Hill Notation):
C9H18O5S
CAS Number:
Molecular Weight:
238.30
PubChem Substance ID:
UNSPSC Code:
12352200
Beilstein/REAXYS Number:
4631
MDL number:
NACRES:
NA.54
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

form

powder

mol wt

238.3

packaging

pkg of 1 g (10724815001), pkg of 5 g (11411446001)

manufacturer/tradename

Roche

shipped in

wet ice

storage temp.

2-8°C

SMILES string

CC(C)S[C@@H]1OC(CO)[C@H](O)C(O)[C@H]1O

InChI

1S/C9H18O5S/c1-4(2)15-9-8(13)7(12)6(11)5(3-10)14-9/h4-13H,3H2,1-2H3/t5-,6+,7+,8-,9+/m1/s1

InChI key

BPHPUYQFMNQIOC-NXRLNHOXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Isopropyl-β-D-thiogalactoside (IPTG) is a chemical analog of galactose that cannot be cleaved by β-galactosidase. IPTG functions by binding to the lacI repressor and altering its conformation, which prevents the repression of the β-galactosidase coding gene lacZ.

Application

IPTG is commonly used in cloning procedures that require induction of β-galactosidase activity. It is used in conjunction with X-Gal or Bluo-Gal in blue-white selection of recombinant bacterial colonies that induce expression of the lac operon in Escherichia coli. IPTG functions by binding to the lacI repressor and altering its conformation, which prevents the repression of the β-galactosidase coding gene lacZ.
Isopropyl-β-D-thiogalactoside (IPTG) is commonly used in cloning procedures that require induction of β-galactosidase activity. It is used in conjunction with X-Gal or Bluo-Gal in blue-white selection of recombinant bacterial colonies that induce expression of the lac operon in Escherichia coli. It has been used for the expression of recombinant genes in E. coli.

Features and Benefits

Assay Time: 50minutes
Specific Activity: The described standard assay will results in a specific activity of 1.8 x 109 dpm/μg, corresponding to 65% incorporation with different substrate DNAs in 30minutes. When varying the ratio of template DNA to labeled dNTP, similar incorporation rates, but different levels of specific activity of the labeled probe are obtained.

Preparation Note

Sample Material
10 ng to 3μg template DNA, linearized and at least 100 to 200bp long
Working concentration: Final concentration should be 0.2 mM.
Storage conditions (working solution): Stock solutions of the product in water (e.g., 1 M IPTG) stored at -15 to -25 °C, are stable up to six months.

Other Notes

For life science research only. Not for use in diagnostic procedures.
Non-metabolizable galactose analog.
The amount of labeled DNA depends on:
  • amount of template DNA
  • purity of template DNA
  • conformation of template DNA
  • average size of labeled fragment

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sonja-Verena Albers et al.
Journal of bacteriology, 185(13), 3918-3925 (2003-06-19)
A large number of secretory proteins in the thermoacidophile Sulfolobus solfataricus are synthesized as a precursor with an unusual leader peptide that resembles bacterial type IV prepilin signal sequences. This set of proteins includes the flagellin subunit but also various
Louis Droogmans et al.
Nucleic acids research, 31(8), 2148-2156 (2003-04-12)
N1-methyladenosine (m1A) is found at position 58 in the T-loop of many tRNAs. In yeast, the formation of this modified nucleoside is catalyzed by the essential tRNA (m1A58) methyltransferase, a tetrameric enzyme that is composed of two types of subunits
L H Hansen et al.
Current microbiology, 36(6), 341-347 (1998-06-03)
The role of the Escherichia coli lacY gene product (the lactose permease) in the induction of isopropyl-beta-D-thiogalactopyranoside (IPTG) inducible promoters was studied in E. coli and P. fluorescens. This was done by comparing strains containing a lacIPOZYA chromosomal insert with

Global Trade Item Number

SKUGTIN
1072481500104061838674692
1141144600104061838701701

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service