Skip to Content
Merck

DTTP-RO

Roche

dTTP

PCR Grade, sodium salt

Synonym(s):

Thymidine 5′-triphosphate sodium salt solution, TTP

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352302
PubChem Substance ID:
MDL number:
NACRES:
NA.54
Assay:
99% (dNTP)
Form:
solution
Storage temp.:
−20°C
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

99% (dNTP)

form

solution

usage

sufficient for 125,000 standard reactions (03732711001), sufficient for 31,250 standard reactions (11969048001), sufficient for 6,250 standard reactions (11934546001)

packaging

pkg of 1,250 μL (11969048001 [100 mM]), pkg of 4 × 1,250 μL (03732711001 [4x100 mM]), pkg of 250 μL (11934546001 [100 mM])

manufacturer/tradename

Roche

concentration

<0.9% (dNDP), 100 mM

storage temp.

−20°C

SMILES string

[Na].CC1=CN(C2CC(O)C(COP(O)(=O)OP(O)(=O)OP(O)(O)=O)O2)C(=O)NC1=O

InChI

1S/C10H17N2O14P3.Na.H/c1-5-3-12(10(15)11-9(5)14)8-2-6(13)7(24-8)4-23-28(19,20)26-29(21,22)25-27(16,17)18;;/h3,6-8,13H,2,4H2,1H3,(H,19,20)(H,21,22)(H,11,14,15)(H2,16,17,18);;

InChI key

KAELIXTVSANDSE-UHFFFAOYSA-N

General description

PCR Grade nucleotides from Roche are specially manufactured and purified to the highest possible chemical purity. PCR Grade nucleotides contain at least 99% of the relevant deoxynucleoside-triphosphate (dNTP) and less than 0.9% deoxynucleoside-diphosphate (dNDP).
Thymidine 5′-triphosphate sodium salt solution is used in DNA synthesis reactions such as PCR, DNA sequencing, cDNA synthesis and molecular cloning techniques.
dTTP, PCR Grade, is a 100 mM clear, colorless solution of the sodium salt (pH 8.3).

Application

dTTP, PCR Grade, is suitable for many applications where high-quality reagents are required. Such procedures include reverse transcription (RT), polymerase chain reaction (PCR), RT-PCR, DNA labeling reactions, and sequencing/cycle sequencing analysis.

Physical form

solution of dTTP in water (pH 8.3).

Preparation Note

Working solution: We recommend to prepare a deoxynucleotide mix containing dATP, dCTP, dGTP, dTTP (10 mM, each); (e.g., for the preparation of 100 μl nucleotide mix add 10 μl of dATP, dCTP, dGTP, dTTP (each) to 60 μl Water, PCR Grade).

Analysis Note

Function tested in RT-PCR. Tested for absence of contaminating deoxyribonucleases, ribonucleases and nicking activities according to the current Quality Control procedures.

Other Notes

For general laboratory use.


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

does not flash

flash_point_c

does not flash



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library