Skip to Content
Merck

DATP-RO

Roche

dATP

99% (dNTP), <0.9% (dNDP), solution, 100 mM

Synonym(s):

2′-Deoxyadenosine 5′-triphosphate sodium salt solution, dATP

Sign In to View Organizational & Contract Pricing.

Select a Size

Pricing and availability is not currently available.

About This Item

UNSPSC Code:
41106300
NACRES:
NA.54

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

dATP, 99% (dNTP), <0.9% (dNDP), solution, 100 mM

InChI

1S/C10H16N5O12P3.Na/c11-9-8-10(13-3-12-9)15(4-14-8)7-1-5(16)6(25-7)2-24-29(20,21)27-30(22,23)26-28(17,18)19;/h3-7,16H,1-2H2,(H,20,21)(H,22,23)(H2,11,12,13)(H2,17,18,19);/q;+1/p-1/t5-,6+,7+;/m0./s1

InChI key

YJWCICGGRLOGEH-VWZUFWLJSA-M

SMILES string

[Na+].Nc1ncnc2n(cnc12)[C@H]3C[C@H](O)[C@@H](COP(O)(=O)OP(O)(=O)OP(O)([O-])=O)O3

description

PCR Grade, sodium salt

assay

99% (dNTP)

form

solution

usage

sufficient for 125,000 standard reactions (03732681001)
sufficient for 31,250 standard reactions (11969013001)
sufficient for 6,250 standard reactions (11934511001)

packaging

pkg of 1,250 μL (11969013001 [100 mM])
pkg of 4 × 1,250 μL (03732681001 [4x100 mM])
pkg of 250 μL (11934511001 [100 mM])

manufacturer/tradename

Roche

concentration

<0.9% (dNDP)
100 mM

storage temp.

−20°C

Quality Level

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
D692011051440001DNSTP-RO
manufacturer/tradename

Roche

manufacturer/tradename

-

manufacturer/tradename

Roche

manufacturer/tradename

Roche

Quality Level

100

Quality Level

200

Quality Level

100

Quality Level

100

assay

99% (dNTP)

assay

≥99%

assay

98%

assay

≥98% (dntp)

form

solution

form

liquid

form

solution

form

liquid

packaging

pkg of 1,250 μL (11969013001 [100 mM]), pkg of 250 μL (11934511001 [100 mM]), pkg of 4 × 1,250 μL (03732681001 [4x100 mM])

packaging

-

packaging

pkg of 250 μL (25 μmol; 100 mM)

packaging

pkg of 4 × 100 μL (11277049001 [4x10 μmol]), pkg of 40 × 100 μL (11922505001 [40x100mM])

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

Application

2′-Deoxyadenosine 5′-triphosphate sodium salt solution is used in DNA sysnthesis reactions such as PCR, DNA sequencing and molecular cloning techniques.
Quality
Function tested in RT-PCR. Tested for absence of contaminating deoxyribonucleases, ribonucleases and nicking activities according to the current Quality Control procedures.
dATP, PCR Grade, is suitable for many applications where high-quality reagents are required. Such procedures include reverse transcription (RT), polymerase chain reaction (PCR), RT-PCR, DNA labeling reactions, and sequencing/cycle sequencing analysis.[1][2]

General description

2′-Deoxyadenosine 5′-triphosphate (dATP) is made up of a nucleobase attached to deoxyribose and a 5′-hydroxyl on the sugar bound to a chain of three phosphate residues. dATP is used by cells for synthesis of DNA by DNA polymerases.
dATP, PCR Grade, is a 100 mM clear colorless solution of the sodium salt (pH 8.3). PCR Grade nucleotides from Roche are specially manufactured and purified to the highest possible chemical purity. PCR Grade nucleotides contain at least 99% of the relevant deoxynucleoside-triphosphate (dNTP) and less than 0.9% deoxynucleoside-diphosphate (dNDP).

Other Notes

For general laboratory use.

Physical form

solution of dATP in water (pH 8.3).

Preparation Note

Working solution: We recommend to prepare a deoxynucleotide mix containing dATP, dCTP, dGTP, dTTP (10 mM, each); (e.g., for the preparation of 100 μl nucleotide mix add 10 μl of dATP, dCTP, dGTP, dTTP (each) to 60 μl Water, PCR Grade).

Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Richard Byers et al.
Diagnostic molecular pathology : the American journal of surgical pathology, part B, 13(3), 144-150 (2004-08-24)
RNA extraction still relies almost exclusively on the use of fresh or frozen tissue, limiting the number of samples that can be analyzed, and there is a growing need for means of global mRNA analysis of archived formalin-fixed paraffin-embedded tissue
Danos C Christodoulou et al.
Current protocols in molecular biology, Chapter 4, Unit4-Unit4 (2011-04-08)
RNA-seq is a method for studying the transcriptome of cells or tissues by massively parallel sequencing of tens of millions of short DNA fragments. However, the broad dynamic range of gene expression levels, which span more than five orders of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service