Skip to Content
Merck

3HBDB-RO

Roche

3-Hydroxybutyrate Dehydrogenase (3-HBDH)

suspension, ~3 units/mg protein (At 25 °C with 3-hydroxybutyrate as the substrate; standardized with BSA.), grade II

Synonym(s):

3-Hydroxybutyrate Dehydrogenase (3-HBDH), 3-HBDH

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352204
EC Number:
NACRES:
NA.54
Specific activity:
~3 units/mg protein (At 25 °C with 3-hydroxybutyrate as the substrate; standardized with BSA.)
Concentration:
5 mg/mL
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

suspension

specific activity

~3 units/mg protein (At 25 °C with 3-hydroxybutyrate as the substrate; standardized with BSA.)

packaging

vial of 2 mL (10127833001), vial of 5 mL (10127841001)

manufacturer/tradename

Roche

concentration

5 mg/mL

impurities

<0.1% LDH, <5% MDH

optimum pH

6.2-6.9(for reduction), 7-9(for oxidation)

shipped in

wet ice

storage temp.

2-8°C

General description

3-Hydroxybutyrate dehydrogenase (3-HBDH) enzyme, obtained from Rhodobacter sphaeroides, is generally used for the quantification of ketone bodies, such as D-3-hydroxybutyrate and acetoacetate.

Application

3-Hydroxybutyrate Dehydrogenase (3-HBDH) oxidizes selectively (R)-3-hydroxymonocarboxylic acids, or reverse reaction.

Biochem/physiol Actions

3-Hydroxybutyrate dehydrogenase (3-HBDH), a mitochondrial enzyme, catalyzes the reversible oxidation of 3-hydroxybutyrate (3HB) to acetoacetate, with NAD as coenzyme. In mammals, acetyl-coA is metabolized, in one of two pathways, to produce acetoacetate and D-3-hydroxybutyrate, which along with acetone are known as ketone bodies. 3-HBDH reversibly reduces this free acetoacetate to D-3-­hydroxybutyrate. In patients with diabetic ketoacidosis (DKA), the ratio of 3HB and acetoacetate can be as high as 10:1, as compared to the normal ratio of 1:1. 3-HBDH enzyme can be used to detect the quantity of 3HB in urine, serum, and blood samples.

Physical form

Suspension in 3.2 M ammonium sulfate solution, pH approximately 6

Preparation Note

Stabilizers: NADH, Ca2+

Analysis Note

Contaminants: <0.1% LDH, <5% MDH

Other Notes

For life science research only. Not for use in diagnostic procedures.


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

No data available

flash_point_c

No data available



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library