Skip to Content
Merck
All Photos(1)

Key Documents

12158167001

Roche

Anti-HA-Biotin, High Affinity (3F10)

from rat IgG1

Synonym(s):

antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rat

Quality Level

conjugate

biotin conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3F10, monoclonal

form

lyophilized (stabilized)

packaging

pkg of 50 μg

manufacturer/tradename

Roche

isotype

IgG1

epitope sequence

YPYDVPDYA

storage temp.

2-8°C

Related Categories

General description

Anti-HA-Biotin, High Affinity (3F10) is a monoclonal antibody for the highly sensitive detection of HA-tagged recombinant proteins, Fab fragments, conjugated to biotin. The Anti-HA-Biotin, High Affinity antibody (clone 3F10) recognizes the same epitope as clone 12CA5. It is a monoclonal antibody whose high affinity and low working concentrations result in less cross-reactivity than with other antibodies to the HA-epitope. Anti-HA-Biotin, High Affinity (3F10) is a biotin conjugate of this clone which is specifically useful in western blotting, ELISA applications and assays using the universal biotin-streptavidin platform, by allowing specific and highly sensitive detection of HA-tagged proteins.

Specificity

Anti-HA-Biotin, High Affinity (3F10) recognizes the 9-amino acid sequence YPYDVPDYA, derived from the human influenza hemagglutinin (HA) protein. This epitope is also recognized in fusion proteins regardless of its position (N-terminal, C-terminal or internal).

Immunogen

Amino acids 98-106 from the human influenza virus hemagglutinin protein

Application

Anti-HA-Biotin, High Affinity (3F10) is used for the detection of HA-tagged recombinant proteins using:
  • Dot blots
  • ELISA (enzyme-linked immunosorbent assay)
  • Western blots
Since Anti-HA High Affinity is a rat monoclonal, it is possible to use it in conjunction with murine monoclonals for double labeling.
It has also been used for immunocytochemistry, immunofluorescence and αScreen format based assay.

Quality

Function test: The Anti-HA-Biotin; High Affinity is function tested by Western blot analysis of a HA-tagged fusion protein.

Preparation Note

Sample Materials
Sample preparation:
Prepare protein extracts containing the HA-tagged protein of interest using any of a variety of standard methods. The following lysis buffers have performed well and should be taken as guidelines:
  • Bacterial extracts: 20 mM Tris, pH 8.0, 100 mM NaCl, cOmplete Protease Inhibitor Cocktail Tablets, followed by freeze-thaw.
  • Mammalian extracts: 50 mM Tris, pH 7.5, 150 mM NaCl, 0.1% Nonidet P40, complete Protease Inhibitor Cocktail Tablets.
  • Other cell lysis buffers may be more appropriate for individual applications. In general, to obtain optimal performance of the affinity matrix:
  • Use protease inhibitors to reduce proteolytic activity. Use complete Protease Inhibitor Cocktail Tablets for most applications.
  • Limit detergent to the lowest concentration levels necessary to obtain adequate cell lysis.
Working concentration: Working concentration of conjugate depends on application and substrate

The following concentrations should be taken as a guideline:
  • Dot blot: 100 ng/ml
  • ELISA: 100 ng/ml
  • Western blot: 100 ng/ml

Reconstitution

Add 1 ml double-distilled water to a final concentration of 50 μg/ml.
Rehydrate for 10 minutes prior to use.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarah Tulin et al.
BMC developmental biology, 10, 83-83 (2010-08-07)
As important regulators of developmental and adult processes in metazoans, Fibroblast Growth Factor (FGF) proteins are potent signaling molecules whose activities must be tightly regulated. FGFs are known to play diverse roles in many processes, including mesoderm induction, branching morphogenesis
Hongying Yang et al.
PloS one, 7(9), e46364-e46364 (2012-10-03)
Chronic inflammation is a major contributing factor in the pathogenesis of many age-associated diseases. One central protein that regulates inflammation is NF-κB, the activity of which is modulated by post-translational modifications as well as by association with co-activator and co-repressor
Yetki Aslan et al.
Development (Cambridge, England), 144(15), 2852-2858 (2017-07-12)
The revolution in CRISPR-mediated genome editing has enabled the mutation and insertion of virtually any DNA sequence, particularly in cell culture where selection can be used to recover relatively rare homologous recombination events. The efficient use of this technology in
Jocelyn F Krey et al.
Nature neuroscience, 16(2), 201-209 (2013-01-15)
L-type voltage gated calcium channels have an important role in neuronal development by promoting dendritic growth and arborization. A point mutation in the gene encoding Ca(V)1.2 causes Timothy syndrome, a neurodevelopmental disorder associated with autism spectrum disorders (ASDs). We report
Yan Hu et al.
Nature communications, 8, 13834-13834 (2017-02-09)
Armadillo repeat containing 5 (ARMC5) is a cytosolic protein with no enzymatic activities. Little is known about its function and mechanisms of action, except that gene mutations are associated with risks of primary macronodular adrenal gland hyperplasia. Here we map

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service