Skip to Content
Merck
All Photos(1)

Key Documents

11426346910

Roche

Anti-Fluorescein-POD, Fab fragments

from sheep

Synonym(s):

antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

sheep

Quality Level

conjugate

peroxidase conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized (stabilized)

packaging

pkg of 150 U

manufacturer/tradename

Roche

storage temp.

2-8°C

General description

Fab fragments from polyclonal anti-fluorescein antibodies, conjugated to horseradish peroxidase. The reagent is an anti-fluorescein antibody, Fab fragments from sheep, conjugated with horse-radishperoxidase (POD). After immunization with fluorescein the sheep IgG (immunoglobulin G) was purified by ion exchange chromatography and the specific IgG was isolated by immunosorption. The Fab fragments obtained by papain digestion were purified by gel filtration, conjugated with POD and stabilized in 60mM Tris-Hepes-buffer, 0.4% bovine immunoglobulin (w/v), 0.2% Germall II (w/v), pH 7.2.

Specificity

The polyclonal antibody reacts with free and bound fluorescein.

Application

Anti-Fluorescein-POD, Fab fragments are used for detection of fluorescein-labeled compounds in:
  • Dot blot
  • ELISA (enzyme-linked immunosorbent assay)
  • Immunohistocytochemistry
  • In situ hybridization
  • Southern blot
  • Western blot

Preparation Note

Working concentration: Working concentration of conjugate depends on application and substrate. The following concentrations should be taken as a guideline:
  • Dot blot: 150 mU/ml
  • ELISA: 50 to 150 mU/ml
  • Immunohistocytochemistry: 250 to 500 mU/ml
  • In situ hybridization: 1.5 to 7.5 U/ml
  • Southern blot: 150 mU/ml
  • Western blot: 500 to 1000 mU/ml

Working solution: 100 mM Tris-HCl, 150 mM NaCl, pH 7.5. If necessary 1% Blocking reagent (w/v), dry milk powder, 1 to 5% heat inactivated fetal calf serum (v/v) or sheep normal serum can be used for reduction of unspecific binding.

Reconstitution

Add 1 ml double-distilled water to a final concentration of 150 U/ml.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alemji A Taku et al.
Development (Cambridge, England), 143(1), 123-132 (2016-01-07)
Olfactory sensory axons target well-defined intermediate targets in the zebrafish olfactory bulb called protoglomeruli well before they form odorant receptor-specific glomeruli. A subset of olfactory sensory neurons are labeled by expression of the or111-7:IRES:GAL4 transgene whose axons terminate in the
Guozhu Ning et al.
Developmental cell, 24(3), 283-295 (2013-02-16)
Craniofacial malformations are common structural birth defects and usually associate with abnormal development of pharyngeal arches. Although some microRNAs have been found to be implicated in chondrogenesis in vitro, few have been shown to be essential for cartilage and bone development
Iku Tsutsui-Kimura et al.
Nature communications, 8, 14304-14304 (2017-02-02)
Impaired motivation is present in a variety of neurological disorders, suggesting that decreased motivation is caused by broad dysfunction of the nervous system across a variety of circuits. Based on evidence that impaired motivation is a major symptom in the
Sandeep Gupta et al.
Developmental biology, 366(2), 125-141 (2012-04-28)
The mammalian hippocampus, a center of neurogenesis in the adult brain, is involved in critical functions such as learning and memory processing. Although there is an overall functional conservation between birds and mammals in the hippocampal region of the brain
Lisa A Thomas et al.
The Journal of comparative neurology, 510(1), 47-67 (2008-07-11)
Cell-cell interactions through adhesion molecules play key roles in the development of the nervous system. Synaptic cell adhesion molecules (SynCAMs) comprise a group of four immunoglobulin (Ig) superfamily members that mediate adhesion and are prominently expressed in the brain. Although

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service