Skip to Content
Merck
All Photos(1)

Key Documents

11243217001

Roche

Laminin

from mouse Engelbreth-Holm-Swarm (EHS) sarcoma

Synonym(s):

Laminin from Engelbreth-Holm-Swarm murine sarcoma basement membrane

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352207

biological source

mouse (Engelbreth-Holm-Swarm (EHS) sarcoma)

Quality Level

sterility

sterile; sterile-filtered

Assay

≥90% (SDS-PAGE)

form

solution

mol wt

900 kDa

packaging

pkg of 1 mg (2 ml)

manufacturer/tradename

Roche

technique(s)

cell culture | mammalian: suitable

UniProt accession no.

storage temp.

−20°C

Gene Information

mouse ... Lamb2(16779)

Looking for similar products? Visit Product Comparison Guide

General description

Laminin is purified as laminin-nidogen complex from mouse Engelbreth-Holm-Swarm (EHS) sarcoma. Laminin is a heterotrimer made from α, β and γ chain subunits. It is a glycoprotein which is required for the structural scaffolding of basement membranes. Laminin supports growth and differentiation of many cell types including epithelial, endothelial, neural, muscle and liver cells.

Biological activity: Tested for the promotion of adherence of HT-1080 cells.
Species specificity: Active on most mammalian cells.

Application

Laminin has been demonstrated to promote the attachment and growth of a variety of cells; used for the coating of culture dishes. It has been used for cell adhesion assay. It is recommended for use as a cell culture substratum at 1-2μg/cm2. The optimal concentration does depend on cell type as well as the application or research objectives.
Laminin supports growth and differentiation of many cell types including epithelial, endothelial, neural, muscle and liver cells. It is recommended for use as a cell culture substratum at 1-2 μg/cm2. The optimal concentration does depend on cell type as well as the application or research objectives.

Quality

Purity: ≥90% (SDS-PAGE) (as laminin nidogen-complex, 1:1)

Sequence

Mouse laminin is composed of three polypeptide chains (A: 400 kD, B1: 230 kD and B2: 220 kD), connected by disulphide bridges, and is glycosylated.

Physical form

Solution, 0.5 mg/ml laminin in 0.15 M NaCl, 2 mM EDTA, 0.05 M Tris-HCl, pH 7.4, sterile
Solution, filtered through 0.2 ?m pore size membrane

Preparation Note

Working concentration: 2 to 5 μg/cm2
2 to 5 μg/cm2 for the coating of cell culture vessels
Storage conditions (working solution): 2 to 8 °C
The undiluted solution is stable at 2 to 8 °C for at least 3 months.
The solution should be thawed carefully at 2 to 8 °C or on ice.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julie V Harness et al.
PloS one, 6(1), e14499-e14499 (2011-01-21)
As human embryonic stem cell (hESC) lines can be derived via multiple means, it is important to determine particular characteristics of individual lines that may dictate the applications to which they are best suited. The objective of this work was
Anindita Dutta et al.
International journal of clinical and experimental pathology, 3(3), 288-302 (2010-03-13)
The interaction of cells with adhesion proteins in the extracellular matrix (ECM) provides signals which affect the morphology, motility, gene expression and survival of adherent cells. In the present communication we cultured K562 cells in presence of fibronectin to study
H Colognato et al.
Developmental dynamics : an official publication of the American Association of Anatomists, 218(2), 213-234 (2000-06-08)
The laminins are a family of glycoproteins that provide an integral part of the structural scaffolding of basement membranes in almost every animal tissue. Each laminin is a heterotrimer assembled from alpha, beta, and gamma chain subunits, secreted and incorporated
John P Snow et al.
Neurobiology of disease, 141, 104881-104881 (2020-04-30)
Alternating hemiplegia of childhood (AHC) is a rare neurodevelopmental disease caused by heterozygous de novo missense mutations in the ATP1A3 gene that encodes the neuronal specific α3 subunit of the Na,K-ATPase (NKA) pump. Mechanisms underlying patient episodes including environmental triggers
Amritha A Candadai et al.
Cells, 10(11) (2021-11-28)
Visual dysfunction resulting from optic neuritis (ON) is one of the most common clinical manifestations of multiple sclerosis (MS), characterized by loss of retinal ganglion cells, thinning of the nerve fiber layer, and inflammation to the optic nerve. Current treatments

Articles

The extracellular matrix (ECM) is secreted by cells and surrounds them in tissues.

The extracellular matrix (ECM) is secreted by cells and surrounds them in tissues.

The extracellular matrix (ECM) is secreted by cells and surrounds them in tissues.

The extracellular matrix (ECM) is secreted by cells and surrounds them in tissues.

See All

Protocols

Coating surfaces with laminin for culturing cells requires specific conditions for optimal results. Protocols for coating coverslips to culture neurospheres and general cell culture are included.

Coating surfaces with laminin for culturing cells requires specific conditions for optimal results. Protocols for coating coverslips to culture neurospheres and general cell culture are included.

Coating surfaces with laminin for culturing cells requires specific conditions for optimal results. Protocols for coating coverslips to culture neurospheres and general cell culture are included.

Coating surfaces with laminin for culturing cells requires specific conditions for optimal results. Protocols for coating coverslips to culture neurospheres and general cell culture are included.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service