11093070910
Roche
Biotin-16-dUTP
Synonym(s):
Bio-16-dUTP tetralithium salt, Biotin-labeled Uridines, 5-(N-[N-Biotinyl-ε-aminocaproyl-γ-aminobutyryl]-3-aminoallyl)-2′-deoxyuridine 5′-triphosphate
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C32H52N7O18P3S
CAS Number:
Molecular Weight:
947.78
MDL number:
UNSPSC Code:
41116100
Recommended Products
form
solution
Quality Level
packaging
pkg of 50 μL (50 nmol; 1mM)
manufacturer/tradename
Roche
color
colorless
solubility
water: miscible
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
General description
Biotin is a water-soluble vitamin which functions as a coenzyme for carboxylases.Biotin-16-dUTP is used to produce biotinylated DNA probes that are commonly used to detect specific DNA and RNA sequences in different methods like Southern and Northern blotting, in situ hybridization, and sandwich assays.
Application
Biotin-16-dUTP has been used:
- For nonradioactive DNA labeling such as random priming, PCR labeling, or nick translation. Biotin-16-dUTP replaces dTTP in the random-primed DNA labeling reaction or nick translation in a ratio of 35% Biotin-16-dUTP and 65% dTTP, as well as in PCR.
- as a substrate for terminal transferase, DNA polymerase I (holoenzyme and Klenow fragment), Taq DNA polymerase, and reverse transcriptase (e.g., Transcriptor). The nucleotide also serves as a substrate for Terminal Transferase in 3′-end labeling
- In fluorescence in situ hybridization (FISH)
Biochem/physiol Actions
Biotin influences gene regulation and chromatin structure by associating with lysine residues in histones. It influences the actions of adaptive immune T and natural killer cells. Deficiency of biotin in mice has indicated enhanced inflammation. Studies have also indicated heightened inflammatory response of human dendritic cells in conditions of biotin deficiency.
Quality
Typical analysis: At least 85% deoxynucleoside triphosphate (Biotin-16-dUTP) (HPLC, area%).
Notes: For your convenience, biotin-dUTP is also available in convenient labeling mixes (single solution with conventional dNTPs and biotin-16-dUTP), and technique-specific mixes (nucleotides, buffers, and enzymes in a single vial).
Notes: For your convenience, biotin-dUTP is also available in convenient labeling mixes (single solution with conventional dNTPs and biotin-16-dUTP), and technique-specific mixes (nucleotides, buffers, and enzymes in a single vial).
Analysis Note
Absorption: The spectral data of Biotin-16-dUTP (at pH 7.3):
Emax241: E = 10.700;
Emax292: E = 7.000;
Emax265: E = 4.400;
A292/265 : 1.6.
Emax241: E = 10.700;
Emax292: E = 7.000;
Emax265: E = 4.400;
A292/265 : 1.6.
Other Notes
Biotin-labeled DNA can be detected with: Streptavidin-alkaline phosphatase conjugate and a chemiluminescent substrate (CSPD, CDP-Star) or a color substrate- Biotin Luminescence Detection Kit.
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
does not flash
Flash Point(C)
does not flash
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Sudhanshu Agrawal et al.
American journal of physiology. Cell physiology, 311(3), C386-C391 (2016-07-15)
The water-soluble biotin (vitamin B7) is indispensable for normal human health. The vitamin acts as a cofactor for five carboxylases that are critical for fatty acid, glucose, and amino acid metabolism. Biotin deficiency is associated with various diseases, and mice
Synthesis in vitro and application of biotinylated DNA probes for human papilloma virus type 16 by utilizing the polymerase chain reaction
DAY PJR
Biochemical Methods, 267, 119-123 (1990)
Di Hu et al.
Journal of neurotrauma, 37(21), 2244-2260 (2020-06-20)
Individuals with spinal cord injury (SCI) often develop debilitating neuropathic pain, which may be driven by neuronal damage and neuroinflammation. We have previously demonstrated that treatment using 670 nm (red) light irradiation alters microglia/macrophage responses and alleviates mechanical hypersensitivity at 7
Zheng Li et al.
Nature, 583(7817), 625-630 (2020-07-17)
The recent discovery of N6-methyladenine (N6-mA) in mammalian genomes suggests that it may serve as an epigenetic regulatory mechanism1. However, the biological role of N6-mA and the molecular pathways that exert its function remain unclear. Here we show that N6-mA
Adjuvant Treatment for POLE Proofreading Domain?Mutant Cancers: Sensitivity to Radiotherapy, Chemotherapy, and Nucleoside Analogues
Van Gool IC, et al.
Clinical Cancer Research, 24(13), 3197?3203-3197?3203 (2018)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service