Skip to Content
Merck

10982962001

Roche

N-Acetyl-β-D-Glucosaminidase Standard (NAG Standard)

from beef kidney

Synonym(s):

NAG standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352204
Specific activity:
~20 mU/mL
Biological source:
bovine kidney
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bovine kidney

form

lyophilized

specific activity

~20 mU/mL

packaging

pkg of 5 × 3 mL (5 x 400 μg)

manufacturer/tradename

Roche

technique(s)

activity assay: suitable

color

white to light yellow

solubility

water: soluble

application(s)

life science and biopharma

storage temp.

20-25°C

General description

Standard for the determination of NAG activity in urine research samples with the aid of the Test-Combination N-acetyl-β-D-glucosaminidase.
N-Acetyl-β-D-Glucosaminidase is a high molecular weight lysosomal enzyme existing as two isofomrs, Isoenzyme-A and Isoenzyme-B. It is present in human kidney. It has high molecular weight. It is highly active in renal proximal tubular cells. It has been reported that overexpressed urine-NAG reflects the proximal tubular dysfunction of the kidney.

Application

The NAG standard is used for the determination of NAG activity in urine in life science research applications, with the aid of N-acetyl-β-D-glucosaminidase. It can be used in automatic analyzers.

Preparation Note

Storage conditions (working solution): 2 to 8 °C
Stability of the standard solution
The solution is stable for at least 1 month stored at 2 to 8 °C, protected from light.
Preparation of the standard solution
Dissolve contents of one bottle with 3 ml double distilled water, close bottle and wait for 5 minutes. Dissolve contents completely by gentle swirling. Avoid creating foam.
Keep protected from light.

Other Notes

For life science research only. Not for use in diagnostic procedures.
Volume Activity: The activity of the NAG standard is approximately 20 mU N-acetyl-β-D-glucosaminidase from beef kidney per ml. For exact values, see data printed on the label.


hcodes

Hazard Classifications

Aquatic Chronic 3

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library