Skip to Content
Merck
All Photos(1)

Key Documents

10783641001

Roche

Polyethylene Glycol 1500

Mr ~1500, pkg of 10 × 4 mL, solution

Synonym(s):

Poly(ethylene glycol), peg 1500, PEG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
12161902
Pricing and availability is not currently available.

form

solution

mol wt

Mr ~1500

packaging

pkg of 10 × 4 mL

manufacturer/tradename

Roche

storage temp.

2-8°C

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(ethylene glycol) (PEG) is a non-ionic hydrophilic polymer and is available in different molecular weights. It helps in the purification and crystal growth of proteins and nucleic acids. PEG and dextran together result in aqueous polymer two phase system, which is required for the purification of biological materials. PEG also interacts with cell membrane, thereby allowing cell fusion.[1][2]

Application

Polyethylene Glycol 1500 (PEG-1500) has been used to mediate cell fusion.[3][4][5]

Physical form

Solution, filtered through 0.2 μm pore size membrane, 50% PEG 1500 (w/v) in 75 mM Hepes (pH 8.0), bottled under nitrogen, ready to use

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Poly(Ethylene Glycol) Chemistry: Biotechnical and Biomedical Applications (2013)
Localization of Pom121 to the inner nuclear membrane is required for an early step of interphase nuclear pore complex assembly.
Funakoshi T, et al.
Molecular Biology of the Cell, 22(7), 1058-1069 (2011)
Usefulness of a non-invasive reporter system for monitoring reprogramming state in pig cells: results of a cell fusion experiment.
Ozawa A, et al.
Journal of Reproduction and Development, 56(4), 363-369 (2010)
Heterokaryon?Based Reprogramming for Pluripotency.
Pereira C F and Fisher A G
Current Protocols in Cell Biology, 4B-1 (2009)
Poly (ethylene glycol) in drug delivery: pros and cons as well as potential alternatives.
Knop K, et al.
Angewandte Chemie (International Edition in English), 49(36), 6288-6308 (2010)

Related Content

Polyethylene glycol (PEG), also sometimes referred to as polyethylene oxide (PEO), is a condensation polymer of ethylene oxide and water that has several chemical properties that make it useful for biological, chemical and pharmaceutical applications.

Polyethylene glycol (PEG), also sometimes referred to as polyethylene oxide (PEO), is a condensation polymer of ethylene oxide and water that has several chemical properties that make it useful for biological, chemical and pharmaceutical applications.

Polyethylene glycol (PEG), also sometimes referred to as polyethylene oxide (PEO), is a condensation polymer of ethylene oxide and water that has several chemical properties that make it useful for biological, chemical and pharmaceutical applications.

Polyethylene glycol (PEG), also sometimes referred to as polyethylene oxide (PEO), is a condensation polymer of ethylene oxide and water that has several chemical properties that make it useful for biological, chemical and pharmaceutical applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service