ZLXLKITUV57
UV Loop Kit
For Milli-Q® HX 7000 SD series water purification systems
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41104200
NACRES:
JA.13
Recommended Products
packaging
pkg of 1 unit
power consumption
57 W
compatibility
for use with Milli-Q® HX
General description
The UV loop kit with a power consumption of 57 W is for use with Milli-Q® HX 7000 SD series water purification systems.
Other Notes
Directions for Use
- Organism Retention: Microorganisms
- Mode of Action: UV irradiation
- Application: General laboratory analysis
- Intended Use: Water purification
- Instructions for Use: This item provides water exposed to bactericidal UV lamp. Refer to system equipment user guide section "Using the System" (USB key in the system box)
- Storage Statement: Store in dry location
- Disposal Statement: Dispose of in accordance with applicable federal, state and local regulations.
Legal Information
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.


Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service