Skip to Content
Merck
All Photos(2)

Key Documents

TGFBMAG-64K-03

Millipore

MILLIPLEX® TGFß Magnetic Bead 3 Plex Kit - Immunology Multiplex Assay

for quantification of TGFß1, TGFß2 and TGFß3 in multiple species

Synonym(s):

3 Plex TGFß Assay, TGFß Magnetic Bead Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.47
Pricing and availability is not currently available.

Quality Level

species reactivity

horse, human, nonhuman primates, feline, pig, mouse, rat, canine

manufacturer/tradename

Milliplex®

assay range

accuracy: 62-128%
sensitivity: 8-12 pg/mL
standard curve range: 9.8-2,500 pg/mL

technique(s)

multiplexing: suitable

compatibility

configured for Premixed

detection method

fluorometric (Luminex xMAP)

shipped in

wet ice

General description

The transforming growth factor beta (TGFβ) system, a superfamily of cytokines as well as signaling pathways, is highly conserved throughout the animal kingdom. TGFβ functions in immune cell development, angiogenesis, wound healing and embryonic development, bone development and plays a critical role in immune response, heart disease, and cancer. In its normal state TGFβ is one of the few classes of proteins able to inhibit cell growth by halting mitosis at the G1 state, inducing cell differentiation or apoptosis. However, during oncogenesis mutations in the TGFβ signaling pathway result in tumor cell resistance to the effects of normally functioning TGFβ, causing proliferation without regulation. Initial research suggests that VE-cadherin may enhance the mutated TGFβ signaling pathway.

The MILLIPLEX® TGFβ 1, 2 and 3 Panel is a 3-plex kit to be used for the simultaneous quantification of the following analytes in serum, plasma and cell/tissue culture samples: TGFβ1, TGFβ2, TGFβ3. The Luminex® xMAP® platform uses a magnetic bead immunoassay format for ideal speed and sensitivity to quantitate multiple analytes simultaneously, dramatically improving productivity while conserving valuable sample volume.

Panel Type: Cytokines/Chemokines

Application

  • Analytes: TGF-β1, TGF-β2, TGF-β3
  • Recommended Species: In addition to human, TGF-β1, TGF-β2, TGF-β3 can be detected in the serum of the following species: mouse, rat, mini pig, hamster, guinea pig, rhesus monkey and cynomolgus monkey samples. Additionally, TGF-β1 and TGF-β2 can be detected in the serum of horse, rabbit and canine. However, the exact amount of cross reactivity has not been determined.
  • NOTE: This assay requires a sample acidification treatment using the provided reagents and protocol.
  • Recommended Sample Types: Serum, plasma, cell/tissue culture supernatants and lysates
  • Recommended Sample Dilution: 25 μL per well of a 1:30 dilution after acidification treatment of serum and plasma samples; cell/tissue culture samples may require dilution prior to acidification treatment (see protocol for details).
  • Assay Run Time: Overnight (16-18 hours) at 2-8°C or alternatively, 2 hours at room temperature (20-25°C).
  • Research Category: Inflammation & Immunology
  • Research Subcategory: Cardiovascular Disease, Chronic Inflammatory Disease, Immune Disorders, Inflammatory Disease, Rheumatoid Arthritis

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Samantha M Logan et al.
PeerJ, 7, e8116-e8116 (2019-11-26)
To conserve energy in times of limited resource availability, particularly during cold winters, hibernators suppress even the most basic of physiologic processes. Breathing rates decrease from 40 breaths/minute to less than 1 breath/min as they decrease body temperature from 37
Lerin R Luckett-Chastain et al.
Experimental dermatology, 26(8), 697-704 (2016-11-29)
It been shown that IL-6 modulates TGF-β1 expression in fibroblasts, however, what role IL-6 plays concerning TGF-βR expression and function in skin is unknown. Therefore, the aim of this study was to investigate the mechanism by which IL-6 might modulates
Lingwen Ying et al.
Diabetes, metabolic syndrome and obesity : targets and therapy, 14, 4661-4670 (2021-12-09)
We aimed to explore the immunological characteristics and cytokine profile of the initial stage of type 1 diabetes. In total, 123 age- and sex-matched subjects with newly diagnosed classic type 1 diabetes mellitus (T1ADM), fulminant type 1 diabetes mellitus (FT1DM)
Inmaculada Parrilla et al.
Animals : an open access journal from MDPI, 10(11) (2020-11-08)
In the context of porcine embryo transfer (ET) technology, understanding the tightly regulated local uterine immune environment is crucial to achieve an adequate interaction between the transferred embryos and the receiving endometrium. However, information is limited on the uterine immune
Yanyuan Wu et al.
Journal of clinical medicine, 11(2) (2022-01-22)
The therapeutic targeting of PD-1/PD-L1 has shown clinical efficacy in treating metastatic breast cancer. We investigated the clinical significance of measuring serum PD-L1 levels in African-American and Hispanic women with breast cancer. PD-L1 levels were measured with the ELISA method

Related Content

Discover benefits of using biomarker detection multiplex immunoassays for companion and agricultural animal health research and find vet med multiplex assays.

Discover benefits of using biomarker detection multiplex immunoassays for companion and agricultural animal health research and find vet med multiplex assays.

Discover benefits of using biomarker detection multiplex immunoassays for companion and agricultural animal health research and find vet med multiplex assays.

Discover benefits of using biomarker detection multiplex immunoassays for companion and agricultural animal health research and find vet med multiplex assays.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service