Skip to Content
Merck

Skip To

SCM007

ReNcell Neural Stem Cell Freezing Medium

ReNcell Neural Stem Cell Freezing Medium is qualified for use with ReNcell immortalized human neural progenitor cell lines, CX.

Synonym(s):

Neural Stem Cell Cryopreservation Culture Media

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352207
NACRES:
NA.75
eCl@ss:
32160801
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

liquid

manufacturer/tradename

Chemicon®

technique(s)

cell culture | stem cell: suitable

input

sample type neural stem cell(s)

shipped in

dry ice

storage temp.

-10 to -25°C

General description

ReNcell Neural Stem Cell Freezing Medium is qualified for use with ReNcell immortalized human neural progenitor cell lines, CX (CHEMICON Catalog No. SCC007) and VM (CHEMICON Catalog No. SCC008) cultured in serum-free conditions with ReNcell NSC Maintenance Medium (CHEMICON Catalog No. SCM005). The optimized formulation allows for consistent cryopreservation and high viability upon thawing and plating.

Application

Cryopreservation of ReNcell immortalized human CX and VM neural stem cell lines in serum-free conditions

Physical form

Serum-free formulation. Contains 10% DMSO

Preparation Note

Store at -20C. Refer to lot expiration date on label.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SCM005SCM003SCM004
description

ReNcell Neural Stem Cell Freezing Medium is qualified for use with ReNcell immortalized human neural progenitor cell lines, CX.

description

ReNcell NSC Maintenance Medium is a defined serum-free, growth factor-free neural stem cell medium that has been optimized for the growth & in vitro differentiation of ReNcell immortalized human neural progenitor cells.

description

The Neural Stem Cell Basal Medium is a defined serum-free, growth factor-free medium that has been optimized for the growth & in vitro differentiation of neural stem cells derived from rodents.

description

The ENStem-A Neural Expansion Medium is a defined serum-free formulation that has been optimized for the culture & expansion of ENStem-A Human Neural Progenitor Cells.

technique(s)

cell culture | stem cell: suitable

technique(s)

cell culture | stem cell: suitable

technique(s)

cell culture | stem cell: suitable

technique(s)

cell culture | stem cell: suitable

form

liquid

form

liquid

form

liquid

form

liquid

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

storage temp.

-10 to -25°C

storage temp.

-10 to -25°C

storage temp.

-10 to -25°C

storage temp.

-10 to -25°C

Quality Level

100

Quality Level

100

Quality Level

300

Quality Level

100

manufacturer/tradename

Chemicon®

manufacturer/tradename

Chemicon®

manufacturer/tradename

Chemicon®

manufacturer/tradename

Chemicon®


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters