Skip to Content
Merck

SCCE009

Normal Human Small Airway Epithelial Cells

Normal Human Small Airway Epithelial Cell line provides an ideal serum-free culture model for the accurate studies of toxicity, cystic fibrosis, asthma, pathogenesis, pharmacology or airway wound healing.

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41106514
NACRES:
NA.81
eCl@ss:
32011203
Biological source:
human epithelium (airway)
Morphology:
(epithelial)
Relevant disease(s):
asthma
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human epithelium (airway)

Quality Segment

packaging

vial of 500,000 cells

manufacturer/tradename

Chemicon®, EpiGRO®, Millipore

morphology

(epithelial)

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

asthma

shipped in

liquid nitrogen

storage temp.

-140 to -196°C

General description

Millipore’s Normal Human Small Airway Epithelial Cell line, when grown in our recommended medium, provides an ideal serum-free culture model for the accurate studies of toxicity, cystic fibrosis, asthma, pathogenesis, pharmacology or airway wound healing. These cells are not exposed to antimicrobials or phenol red when cultured in EpiGRO SAE Complete Media Kit. Normal Human Small Airway Epithelial Cells are cryopreserved at passage 1 to ensure highest viability and plating efficiency.

This product is for Research Use Only. This product is not approved for human or veterinary use, or for use in in vitro diagnostic or clinical procedures
Product Source: Human Small Airway Epithelia

Application

Please see data sheet for detailed directions on Thawing, Plating , Feeding, Passaging, etc.
Research Category
Stem Cell Research

Biochem/physiol Actions

Airway
Epithelial Cells

Packaging

500,000 cells

Preparation Note

To maintain the cells’ integrity, Millipore recommends unpacking the products immediately upon receipt and storing at a temperature lower than -150°C or in liquid nitrogen vapor phase.

Analysis Note

Cells are negative for HIV (1,2), HBV, HCV, bacteria, mycoplasma and
fungal contamination.

Further growth: Guaranteed to provide 15 or more doublings when grown in the appropriate EpiGRO Medium.

Other Notes

SCCE009

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
EPIGRO is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


flash_point_f

Not applicable

flash_point_c

Not applicable

Storage Class

12 - Non Combustible Liquids

wgk

WGK 3



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library