Skip to Content
Merck

SCC282

SF188 Human Glioblastoma Cell Line

SF188 human glioblastoma cell line is an important cellular model for pediatric human glioblastoma.

Synonym(s):

SF-188 cells

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.81
UNSPSC Code:
41106514
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


application(s)

cell analysis

General description

Glioblastoma multiforme is the most common and malignant primary brain tumor, with a high recurrence rate and a five-year survival rate of less than 5% (1). Glioblastomas represent approximately 10% of brain tumors in children and pediatric forms often have characteristics distinct from adult forms (2). Glioblastomas are the most aggressive form of cancer, highlighting the importance of relevant human patient-derived cell models for advancing research into characteristics and treatment of this disease.

SF188 is a patient-derived glioblastoma cell line originating from a temporal lobe tumor (3). SF188 cells do not express the glial markers GFAP or glutamine synthetase but are positive for laminin and fibronectin expression in early passages, suggestive of proliferative or transformed mesenchymal cells of glioblastoma (3). SF188 cells exhibit hypertriploidy and epithelial morphology in culture and are non-tumorigenic in athymic mice (3). The SF188 cell line possesses genetic amplification of the proliferative factor c-myc (4).

<bold>Source:</bold>
The SF188 glioblastoma cell line was derived from the left temporal lobe tumor of an 8-year-old male patient (3).

Research Category:
Cancer

Biochem/physiol Actions

Human, Cancer Cells

Packaging

&ge;1X106 cells/vial

Preparation Note

Store in liquid nitrogen. The cells can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.

Other Notes

Subject to local law, this product is intended to be sold for internal in vitro research use only subject to terms and conditions found here: www.sigmaaldrich.com/restrictedcelluse. This product may not be: re-engineered or copied; used to make derivatives, modifications or functional equivalents; used to obtain patents or other IP claiming use of the product; used to develop, test, or manufacturer a commercial product; used as a component in a commercial product; resold or licensed; used in any clinical applications or trials; or used in humans. A license or limited commercial use agreement is required for use by any for-profit entity, use in services, and use in sponsored academic research. For information regarding any such use, please contact licensing@milliporesigma.com.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library