Skip to Content

SCC205

LUVA Human Mast Cell Line

Synonym(s):

Laboratory of University of VirginiA Cell Line

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.81
UNSPSC Code:
41106514
Biological source:
human
Technique(s):
cell culture | mammalian: suitable
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human

Quality Segment

packaging

vial of ≥1X10⁶ cells/vial vial

manufacturer/tradename

Millipore

technique(s)

cell culture | mammalian: suitable

shipped in

liquid nitrogen

storage temp.

−196°C

General description

Mast cells are a type of granulocyte that are important components for the immune system and the process of inflammation. When activated, they will release compounds inducing an inflammatory response and anaphylaxis. They are also directly involved in causing allergies and related disorders. Previously, mast cell lines such as LAD2 were used as in vitro cell models for studying mast cells. However, they were unstable and inconsistent for culturing.1A new mast cell line was developed and given the name of LUVA cells, which mirror the morphology and functionality of human mast cells.

The CD34+ mast cells were cultured for 8 weeks with stem cell factor and cultured with IL-6 and IL-3 for only one week. The stable LUVA cells were maintained and became capable of growing without further addition of growth factors in culture, making them highly convenient for culturing. They are the first mast cell line in which the donor patient did not have a mast cell disorder, as well as having no mutation in the c-kit receptor.1

The LUVA cell line allows for a proper in vitro mast cell model in which researchers can study the mechanisms which cause mastocytosis, a condition where mast cells aggregate in certain parts of the body.2 This condition results in a higher risk of life-threatening allergic reactions as well as rarely resulting in mast cell cancers. Commonly, mastocytosis is caused by a mutation of the c-KIT protein, a receptor tyrosine kinase. By understanding c-KIT mechanisms, therapies can continue to develop and improve for mastocytosis patients.

Source: The LUVA mast cell line was derived from CD34+ cells of a non-mastocytosis donor with aspirin-exacerbated respiratory disease.

References
1. Laidlaw, T.M., et al. Characterization of a novel human mast cell line that responds to stem cell factor and expresses functional FcεRI. Journal of Allergy and Clinical Immunology 127:815-822.e5 (2011)

Application

  • Each vial contains >1X106 viable cells.
  • Cells are tested negative for infectious diseases by a Mouse Essential CLEAR Panel by Charles River Animal Diagnostic Services.
  • Cells are verified to be of human origin and negative for interspecies contamination from human, rat, Chinese hamster, Golden Syrian hamster, and nonhuman primate (NHP) as assessed by a Contamination Clear panel by Charles River Animal Diagnostic Services
  • Cells are negative for mycoplasma contamination.

Features and Benefits

The LUVA cell line is a human mast cell in vitro cell model used to study the mechanisms which cause mastocytosis, inflamation and allergic responses.

Preparation Note

LUVA human mast cells should be stored in liquid nitrogen. The cells can be cultured for at least 10 passages without significantly affecting cell marker expression and function.

Other Notes

Subject to local law, this product is intended to be sold for internal in vitro research use only subject to terms and conditions found here: www.sigmaaldrich.com/restrictedcelluse. This product may not be: re-engineered or copied; used to make derivatives, modifications or functional equivalents; used to obtain patents or other IP claiming use of the product; used to develop, test, or manufacturer a commercial product; used as a component in a commercial product; resold or licensed; used in any clinical applications or trials; or used in humans. A license or limited commercial use agreement is required for use by any for-profit entity, use in services, and use in sponsored academic research. For information regarding any such use, please contact licensing@milliporesigma.com.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Storage Class

12 - Non Combustible Liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library