Skip to Content
Merck

RSB

READYBLUE® PROTEIN GEL STAIN

Synonym(s):

protein gel stain, protein stain

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.32
UNSPSC Code:
41105322
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


quality

MQ100

Quality Level

technique(s)

protein staining: suitable

General description

ReadyBlue® Protein Gel Stain is a rapid and sensitive colloidal Coomassie stain for polyacrylamide gels. The stain is provided as a ready-to-use solution, allowing for a fast and simplified protocol. No prior fixing step is necessary.
  • Colorimetric or IR fluorescence (800 nm) imaging
  • Reusable up to 3 times
  • No organic solvents or phosphoric acid
  • Compatible with mass spectrometry
  • Room temperature storage—it′s ready when you are

Application

Protein gel stain

Preparation Note

Store ReadyBlue® Protein Gel Stain at room temperature. Protect From Light.

Legal Information

READYBLUE is a registered trademark of Merck KGaA, Darmstadt, Germany




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
RSB-30ML04061841447351
RSB-1L04061841447344