Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
41106509
NACRES:
NA.81
eCl@ss:
32011203
Biological source:
mouse
biological source
mouse
Quality Segment
manufacturer/tradename
Specialty Media, EmbryoMax®
technique(s)
cell culture | stem cell: suitable
input
sample type primary embryotic fibroblasts (PMEFs)
sample type induced pluripotent stem cell(s)
sample type: human embryonic stem cell(s)
sample type: mouse embryonic stem cell(s)
shipped in
liquid nitrogen
storage temp.
-140 to -196°C
General description
The EmbryoMax range of PMEF cells provides researchers with a convenient solution for ES cell culture by eliminating the need for time consuming feeder cell isolation and preparation. Many embryonic stem cell culture protocols necessitate the use of primary mouse embryo fibroblast (PMEF) cells. In these protocols, ES cells are typically cultured on a monolayer of PMEF feeder cells. Feeder cells perform two important roles in stem cell culture: they secrete several important growth factors into the medium, which help maintain pluripotency, and they provide a cellular matrix for ES cells to grow.
These PMEF cells were isolated from CF-1 mouse embryos at day 13 and mitotically inactivated by treatment with irradiation. The cells can be directly plated onto gelatin coated (Cat. No. ES-006-B) culture vessels. These cells are from passage 3. 5-6 x 106 cells will produce the appropriate density feeder layer on the following tissue culture vessels:
• 6 x 60mm dishes
• 2 x T75 flasks
• 2 x 6 well plates
Refer to Murine Embryonic Stem Cell Culture Manual for complete culture protocols and media formulations for PMEF cells.
• 6 x 60mm dishes
• 2 x T75 flasks
• 2 x 6 well plates
Refer to Murine Embryonic Stem Cell Culture Manual for complete culture protocols and media formulations for PMEF cells.
Application
Research Category
Stem Cell Research
Stem Cell Research
Biochem/physiol Actions
Primary Mouse Embryo Fibroblasts
Packaging
5-6x106 ea
Preparation Note
Vials should be stored in the vapor phase of liquid nitrogen.
Legal Information
EmbryoMax is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Still not finding the right product?
Explore all of our products under EmbryoMax® Primary Mouse Embryonic Fibroblasts
Storage Class
10 - Combustible liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.