Skip to Content
Merck

PHCC3WKIT

Scepter 3.0 Mounting Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NC.57
UNSPSC Code:
41131501
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

pkg of 1 ea

manufacturer/tradename

Scepter

technique(s)

cell counting: suitable

compatibility

Scepter

shipped in

ambient

storage temp.

no temp limit

General description

The Scepter 3.0 Charging Station Mounting Kit is used to mount the the Scepter 3.0 Charging Station (sold separately) in multiple mounting options, freeing up valuable hood or lab bench space.

A Charging Station Mounting Kit is included with Scepter 3.0 instruments and can also be purchased separately for mulitple mounting options.

Other Notes

Included with Scepter 3.0 instruments:PHCC340KITPHCC360KITCan be purchased separately for additional mounting options.




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library