Skip to Content
Merck
All Photos(1)

Documents

PC138

Sigma-Aldrich

Anti-NF-κB (p65, RelA) (Ab-2) Rabbit pAb

liquid, Calbiochem®

Synonym(s):

Anti-Nuclear Factor ΚB

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Related Categories

General description

Rabbit polyclonal antibody. Recognizes the ~65 kDa NF-κB p65 (RelA) protein.
Recognizes the ~65 kDa NF-κB p65 (RelA) protein in Daudi cells.
This Anti-NF-κB (p65, RelA) (Ab-2) Rabbit pAb is validated for use in ELISA, Gel Shift, Immunoblotting, Immunoprecipitation for the detection of NF-κB (p65, RelA) (Ab-2).

Immunogen

Human
a synthetic peptide corresponding to amino acids near the N-terminal domain of human NF-κB p65 (RelA), conjugated to KLH

Application

ELISA (see comments)

Gel Shift (see comments)

Immunoblotting (1:500, chemiluminescence)

Immunoprecipitation (see comments)

Warning

Toxicity: Standard Handling (A)

Physical form

In 5 mM sodium phosphate buffer, pH 7.4.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Positive Control
Daudi cells

Other Notes

Kolberg, R., 1997. J. NIH Research9, 29.
Van Antwerp, D.J. et al. 1996. Science274, 787.
Beg, A.A., et al. 1995. Nature376, 167.
Baeuerle, P.A., and Henkel, T., 1994. Ann. Rev. Immunol.12, 141.
Wulczyn, F.G., et al. 1992. Nature358, 597.
This antibody has also been reported to work for ELISA, gel shift, and immunoprecipitation. Antibody should be titrated for optimal results in individual systems.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chiara Riganti et al.
American journal of respiratory cell and molecular biology, 36(6), 746-756 (2007-02-27)
We have observed that in three human malignant mesothelioma cell lines, crocidolite asbestos induced the activation of the transcription factor NF-kappaB and the synthesis of nitric oxide (NO) by inhibiting the RhoA signaling pathway. The incubation with crocidolite decreased the
Cielo Mae D Marquez et al.
Asian Pacific journal of cancer prevention : APJCP, 21(8), 2453-2461 (2020-08-29)
The chemotherapy drug doxorubicin (DOX) is effective in treating many types of cancers. However, due to its pro-inflammatory and cardiotoxic side effects, other remedies have also been explored as alternative treatments. The plant Alangium longiflorum was reported to contain cytotoxic
Gaoxiang Huang et al.
Cellular & molecular immunology, 14(3), 265-275 (2015-09-22)
Immune cells, particularly macrophages, play critical roles in the hypoxia-induced inflammatory response. The small GTPase RhoB is usually rapidly induced by a variety of stimuli and has been described as an important regulator of cytoskeletal organization and vesicle and membrane
Raquel Fernández-Ginés et al.
Redox biology, 55, 102396-102396 (2022-07-16)
It is widely accepted that activating the transcription factor NRF2 will blast the physiological anti-inflammatory mechanisms, which will help combat pathologic inflammation. Much effort is being put in inhibiting the main NRF2 repressor, KEAP1, with either electrophilic small molecules or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service