Skip to Content

OP145

Anti-MDM2 (Ab-5) Mouse mAb (4B2C1.11)

liquid, clone 4B2C1.11, Calbiochem®

Synonym(s):

Anti-Ubiquitin Protein Ligase, Anti-p53 Binding Protein, Ant-Murine Double Minute Chromosome-2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.43
UNSPSC Code:
12352203
Clone:
4B2C1.11, monoclonal
Species reactivity:
human
Application:
Citations:
6
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

antibody form

purified antibody

antibody product type

primary antibodies

clone

4B2C1.11, monoclonal

form

liquid

does not contain

preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, avoid repeated freeze/thaw cycles

dilution

(Immunoblotting (2 µg/mL, chemiluminescence)
Immunofluorescence (2.5 µg/mL)
Immunoprecipitation (1 µg/reaction,
Paraffin Sections (2.5 µg/mL, heat pre-treatment required))

isotype

IgG1

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MDM2(4193)

General description

Purified mouse monoclonal antibody. Recognizes the ~90 kDa (apparent MW) MDM2 protein.
Recognizes the ~90 kDa (apparent MW) MDM2 protein in A549 and MCF-7 cells and breast carcinoma tissue.
This Anti-MDM2 (Ab-5) Mouse mAb (4B2C1.11) is validated for use in Immunoblotting, Immunofluorescence, Immunoprecipitation, Paraffin Sections for the detection of MDM2 (Ab-5).

Immunogen

Human
recombinant, human MDM2

Application


Immunoblotting (2 g/ml, chemiluminescence)
Immunofluorescence (2.5 g/ml)
Immunoprecipitation (1 g/reaction, see application references)
Paraffin Sections (2.5 g/ml, heat pre-treatment required)

Packaging

Please refer to vial label for lot-specific concentration.

Physical form

In 50 mM sodium phosphate buffer, 50% glycerol, pH 7.5.

Preparation Note

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Positive Control
A549 or MCF7 cells or breast carcinoma tissue

Other Notes

Antibody should be titrated for optimal results in individual systems.
Marchetti, A., et al. 1995. J. Pathol.175, 31.
Barak, Y., et al. 1993. EMBO. J.12, 461.
Ladanyi, M., et al. 1993. Cancer Res.53, 16.
Leach, F.S., et al. 1993. Cancer Res.53, 2231.
Oliner, J.D., et al. 1993. Nature362, 857.
Momand, J., et al. 1992. Cell69, 1237.
Oliner, J.D., et al. 1992. Nature358, 80.
Fakharzadeh, S.S., et al. 1991. EMBO J.10, 1565.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library