NI02
Normal Goat IgG
This Normal Goat IgG is validated for use in ELISA Flow Cytometry Immunoblotting Immunofluorescence Immunohistochemistry Immunoprecipitation for the detection of Goat IgG, Non-immune.
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
NACRES:
NA.43
Recommended Products
biological source
goat
Quality Level
form
liquid
contains
≤0.1% sodium azide as preservative
manufacturer/tradename
Calbiochem®
storage condition
do not freeze
shipped in
wet ice
storage temp.
2-8°C
General description
Native IgG from goat serum. IgG is a glycoprotein found in serum and tissue fluids. For use as a negative control in parallel with specific goat IgG-type primary antibodies.
Application
ELISA
Flow Cytometry
Immunoblotting
Immunofluorescence
Immunohistochemistry
Immunoprecipitation
Flow Cytometry
Immunoblotting
Immunofluorescence
Immunohistochemistry
Immunoprecipitation
Warning
Toxicity: Standard Handling (A)
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Makoto Kondo et al.
NPJ Regenerative medicine, 6(1), 65-65 (2021-10-17)
Knee cartilage does not regenerate spontaneously after injury, and a gold standard regenerative treatment algorithm has not been established. This study demonstrates preclinical safety and efficacy of scaffold-free, human juvenile cartilage-derived-chondrocyte (JCC) sheets produced from routine surgical discards using thermo-responsive
Hiroai Okutani et al.
Glia, 66(8), 1775-1787 (2018-04-26)
Glial cells play important roles in the development and maintenance of neuropathic pain. In particular, activated microglia in the spinal cord facilitate the hyper-excitability of dorsal horn neurons after peripheral nerve injury via pro-inflammatory molecules. In this study, we investigated
Bernhard Gesslbauer et al.
Annals of neurology, 82(3), 396-408 (2017-08-24)
Axons traveling within the brachial plexus are responsible for the dexterous control of human arm and hand movements. Despite comprehensive knowledge on the topographical anatomy of nerves innervating the human upper limbs, the definite quantity of sensory and motor axons
Kevin Wilhelmsen et al.
Innate immunity, 18(4), 602-616 (2011-12-22)
The vascular endothelium is integrally involved in the host response to infection and in organ failure during acute inflammatory disorders such as sepsis. Gram-negative and Gram-positive bacterial lipoproteins circulate in sepsis and can directly activate the endothelium by binding to
Malgorzata Grzesiak et al.
Histochemistry and cell biology, 155(1), 101-116 (2020-10-24)
Vitamin D3 (VD3) plays an important role in the ovary and its deficiency is associated with ovarian pathologies, including polycystic ovary syndrome (PCOS). However, there is no data related to VD3 metabolism in the ovary during PCOS. Herein, we investigated
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service