Select a Size
About This Item
Product Name
Methanol, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS
Quality Level
grade
ACS reagent, reagent grade
agency
meets USP/NF
vapor density
1.11 (vs air)
vapor pressure
128 hPa ( 20 °C), 410 mmHg ( 50 °C), 97.68 mmHg ( 20 °C)
description
for analytical purposes, for extraction
assay
≥99.8% (GC)
form
liquid
autoignition temp.
725 °F
expl. lim.
36 %
dilution
(for analytical testing)
impurities
≤0.0003 meq/g Titrable acid, ≤0.001% Carbonyl (acetone, formaldehyde, acetaldehyde), ≤0.1% Water (H2O), ≤2 ppm Benzene (GC), 0.0002 meq Titrable base
evapn. residue
≤0.001%
color
clearAPHA: ≤10
refractive index
n20/D 1.329 (lit.)
bp
64.7 °C (lit.)
mp
−98 °C (lit.)
transition temp
flash point 9.7 °C
density
0.791 g/mL at 25 °C (lit.)
application(s)
environmental
food and beverages
industrial qc
pharmaceutical
sample preparation
format
neat
shipped in
ambient
storage temp.
room temp
SMILES string
CO
InChI
1S/CH4O/c1-2/h2H,1H3
InChI key
OKKJLVBELUTLKV-UHFFFAOYSA-N
General description
Exceptional Quality Standards: These solvents meet or exceed ACS specifications, providing assurance of high quality that is crucial for accurate testing and analysis in quality control.
Consistent Reproducibility: Designed for precision, the solvents ensure that results can be reliably reproduced, which is vital for maintaining compliance and integrity in quality control processes.
Versatile Applications: Suitable for a variety of QC tasks, including analytical testing, method validation, and testing USP/NF monographs, these solvents support a comprehensive approach to quality assurance in laboratory settings.
Application
Packaging
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!
Analysis Note
Still not finding the right product?
Explore all of our products under Methanol
signalword
Danger
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
target_organs
Eyes,Central nervous system
Storage Class
3 - Flammable liquids
wgk
WGK 2
flash_point_f
49.5 °F - closed cup
flash_point_c
9.7 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| MX0485P-4 | 04061839429147 |
| MX0485-5 | 04061839428980 |
| MX0485-3 | 04061839428898 |
| MX0485-20 | 04061839428881 |
| MX0485-9 | 04061839429024 |
| MX0485-31 | 04061839428904 |
| MX0485-7 | 04061839429000 |
| MX0485-37 | 04061839428942 |
| MX0485-6 | 04061839428997 |
| MX0485-8 | 04061839429017 |


