MSPNNVX00
Multiscreen® 96 well Plate, solid bottom
V-bottom natural polypropylene plate, non-sterile, pkg of 100 plates
Synonym(s):
MultiScreen® 96-well plate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122102
NACRES:
NB.15
material:
V-bottom natural polypropylene plate
size:
96 well
sterility:
non-sterile
Recommended Products
material
V-bottom natural polypropylene plate
Quality Level
sterility
non-sterile
product line
MultiScreen®
packaging
pkg of 100 plates
plate size
96 wells
size
96 well
Looking for similar products? Visit Product Comparison Guide
General description
MultiScreen® 96-well polypropylene microplates meet industry standard dimensions for use in common biochemical assay platforms, including automated liquid handling, microplate readers, and other microplate compatible devices. Polypropylene microplates are appropriate for sample preparation and storage in addition to some biochemical assays.
Physical form
Color Code:Natural
Other Notes
Automation Compatible:Yes
Legal Information
MULTISCREEN is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service