Skip to Content
Merck
All Photos(5)

Key Documents

MMHEADMM2

Millipore

Milliflex Oasis® Filtration Head

for use with Milliflex Oasis® filtration units, designed for MMSYSTMM2 pump

Synonym(s):

Filtration Accessory, Filtration Head, Milliflex Oasis® filtration head, Milliflex Oasis® filtration pump head

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NB.79

material

316 stainless steel (ASI)

Quality Level

packaging

pkg of 1 unit

manufacturer/tradename

Milliflex Oasis®

application(s)

bioburden testing
food and beverages
pharmaceutical
water monitoring

compatibility

for use with Milliflex Oasis® filtration units, designed for MMSYSTMM2 pump

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Milliflex Oasis® filtration system offers a solution to simplify and secure microbial testing process with premium levels of productivity, result reliability and regulatory compliance.
The Milliflex Oasis® filtration head is made of 316L stainless steel and can be autoclaved at 121°C for up to 30 minutes or at 134°C for up to 15 minutes.
The Milliflex Oasis® filtration head has been designed to fit the Milliflex Oasis® vacuum pump MMSYSTMM2 and is also compatible with the Milliflex Oasis® vacuum pump, VHP resistant, MMPPLUVHP.

Application

Milliflex Oasis® Filtration Head is used for filtration of liquid samples for Bioburden testing, water monitoring, and general microbiological analysis of various filtrable fluids.

Legal Information

MILLIFLEX OASIS is a registered trademark of Merck KGaA, Darmstadt, Germany

related product


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Milliflex® Quantum system enables rapid microbial contamination detection with fluorescence staining technology.

Milliflex® Quantum system enables rapid microbial contamination detection with fluorescence staining technology.

Milliflex® Quantum system enables rapid microbial contamination detection with fluorescence staining technology.

Milliflex Oasis® system monitors bioburden in various samples, increasing lab productivity.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service