Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.79
UNSPSC Code:
41116129
material
silicone
Quality Segment
packaging
pkg of 5 units
manufacturer/tradename
Milliflex Oasis®
application(s)
cosmetics
food and beverages
pharmaceutical
water monitoring
compatibility
for use with Milliflex Oasis® system
shipped in
ambient
General description
Milliflex Oasis® internal flow sanitization gasket kit is an accessory for the filtration system. Milliflex Oasis® filtration system offers a solution to simplify and secure microbial testing process with premium levels of productivity, result reliability and regulatory compliance. The pump is a new generation vacuum pump for water and bioburden analysis in pharmaceutical environments. The Milliflex Oasis® sanitization kit is used to decontaminate the interior of the pump. The sanitization spare part kit allows to maintain the Milliflex Oasis® internal flow sanitization kit when requested.
Application
Milliflex Oasis® filtration pump finds its application in bioburden testing and water monitoring of beer, bottled water, cosmetics and Pharmaceutical Products Analysis
Other Notes
- Five U-rings for funnel-to-kit tightness
- Five O-rings for kit-to-pump tightness
Legal Information
MILLIFLEX OASIS is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| MMGASKTSK | 04061839792838 |