Skip to Content
Merck

MLID1

mPAGE® Tank Lid with Electrode Cables

replacement part for mPAGE® Mini Gel Tank and mPAGE® Wet Transfer System

Synonym(s):

electroporesis lid, lid with power cables

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NC.01
UNSPSC Code:
41105305
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


AC/DC input

100 watts (maximum power limit), 300 V (maximum voltage limit)

packaging

pkg of 1 pack

parameter

70 °C ambient temp. limit

technique(s)

SDS-PAGE: suitable, electrophoresis: suitable

compatibility

for use with mPAGE® Mini Gel Tank

General description

mPAGE® Tank Lid with Electrode Cables for use with mPAGE® Mini Gel Tank and mPAGE® Mini Wet Transfer SystemThe mPAGE® Mini Gel Tank is used to perform high-resolution protein separation on handcast or precast polyacrylamide gels. The gel tank is designed for use with 8 cm x 10 cm mini gels and is compatible with gels cast on the mPAGE® Gel Caster and mPAGE® Precast Gels. The mPAGE® Mini Gel Tank is available in 2 formats for running up to 2 or 4 mini gels and is designed to prevent incorrect assembly and leakage.
The mPAGE® Mini Wet Transfer System (MWTS) accommodates two gel holder cassettes for electrophoretic transfer of proteins from mini gels to PVDF or nitrocellulose blotting membrane using mPAGE® Transfer Buffer or a compatible transfer buffer.

Legal Information

MPAGE is a registered trademark of Merck KGaA, Darmstadt, Germany




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library