MHA00P2TT
Tryptone Glucose Extract Broth
with triphenyl tetrazolium chloride, ready-to-use, suitable for microbiology, for heterotrophic bacteria
Synonym(s):
TGE broth with TTC, TGE broth with triphenyl tetrazolium chloride
About This Item
Recommended Products
sterility
sterile
Quality Level
form
liquid
shelf life
12 mo.
feature
ready-to-use
composition
with triphenyl tetrazolium chloride
packaging
pack of 50 tests
technique(s)
cell counting: suitable (total viable count (TVC))
capacity
2 mL
filling volume
2 mL
color
clear medium amberslightly opalescent
input
sample type beverage(s)
sample type food(s)
sample type cider
sample type bottled water
sample type beverage(s)
sample type beer
sample type cosmetics
sample type wine
sample type sport drink
sample type soft drink
pH
7.0±0.2 (25.0 °C)
application(s)
bioburden testing
clinical
cosmetics
environmental
food and beverages
industrial qc (microbial monitoring)
water monitoring
microbiology
compatibility
for use with MF-method
for use with Microfil®
for use with MicropreSure®
for use with Milliflex®
shipped in
wet ice
storage temp.
2-8°C
suitability
heterotrophic bacteria
nonselective and differential for
Related Categories
General description
Application
Features and Benefits
- Ready-to-use Liquid ampoule media solve numerous problems in microbiology laboratories where time, equipment, and trained personnel are often in short supply.
- They are available in convenient, single-test 2 mL plastic ampoules, eliminating the manipulation of glass ampoules and avoiding the waste and spoilage associated with bulk packaged media.
Specifications
Analysis Note
E. coli (ATCC 25922)
Legal Information
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service