Skip to Content
Merck

MABS436

Anti-ATE1 Antibody, clone 6F11

clone 6F11, from rat

Synonym(s):

Arginyl-tRNA--protein transferase 1, Arginyltransferase 1, R-transferase 1, Arginine-tRNA--protein transferase 1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
6F11, monoclonal
Application:
ICC, WB
Citations:
9
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rat

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6F11, monoclonal

species reactivity

human, mouse

technique(s)

immunocytochemistry: suitable, western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ATE1(11101)

General description

ATE1 plays an important role a specialized ubiquitin mediated protein degradation pathway. ATE1 is involved in the post-translational conjugation of arginine to the N-terminal or mid-chain Aspartate or Glutamate amino acids of proteins. The arginylation is required for effective ubiquitin mediated protein degradation. ATE1 is a central player in the Arg/N-end rule pathway of ubiquitin mediated protein degradation. This system disposes of malformed or misdirected proteins and protein fragments. ATE1 has two isoforms, isoform 1 is nuclear localized and isoform 2 is cytoplasmically localized. ATE1 is widely expressed, and recent research demonstrates that ATE1 mediated arginylation is essential for platelet myosin phosphorylation, clot retraction and thrombosis formation. Recent research also shows that the Arg/N-end rule pathway and ATE1 may play a role in helping to prevent neurodegeneration caused by the buildup of protein fragments found in various neurological disease states such as Alzheimer′s disease, amyotrophic lateral sclerosis, and Parkinson′s disease.
~60 kDa observed

Immunogen

Recombinant protein corresponding to mouse ATE1.

Application

Research Category
Signaling
Research Sub Category
Signaling Neuroscience
This Anti-ATE1 Antibody, clone 6F11 is validated for use in Western Blotting and Immunocytochemistry for the detection of ATE1.
Western Blotting Analysis: 0.5 µg/mL from a representative lot detected ATE1 in 10 µg of mouse liver tissue lysate.
Western Blotting Analysis: A representative lot detected ATE1 in ATE1 isoforms and mouse liver extract tissue lysate (Wang, J., et al. (2011). Chemistry & Biology. 18:121-130).
Immunocytochemistry Analysis: A representative lot detected ATE1 in immunostaining and cell extract preparation, immortalized mouse embryonic fibroblasts derived from wild-type and Ate1 cells (Wang, J., et al. (2011). Chemistry & Biology. 18:121-130).

Physical form

Format: Purified
Protein G Purified
Purified rat monoclonal IgG2bκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in HepG2 cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected ATE1 in 10 µg of HepG2 cell lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library