Skip to Content
Merck
All Photos(4)

Documents

MABN471

Sigma-Aldrich

Anti-Tau Antibody, PAD, clone TNT-1

clone TNT-1, from mouse

Synonym(s):

Microtubule-associated protein tau, Neurofibrillary tangle protein, Paired helical filament-tau, PHF-tau, Tau

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

TNT-1, monoclonal

species reactivity

mouse, rat, human

technique(s)

ELISA: suitable
dot blot: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MAPT(4137)
mouse ... Mapt(17762)
rat ... Mapt(29477)

Related Categories

General description

Tau or Microtubule-associated protein tau (MAPT), also known as neurofibrillary tangle protein and paired helical filament-tau (PHF-tau), is a cytosolic protein that promotes microtubule assembly and stability, and might be involved in the establishment and maintenance of axonal polarity in neurons. Tau binds to and is thought to function as a linker protein between axonal microtubules and neural plasma membrane components. There are multiple isoforms, and the short isoforms allow plasticity of the cytoskeleton whereas the longer isoforms may preferentially play a role in its stabilization. PAD is the phosphatase activating domain, and has been demonstrated to be involved in the inhibition of anterograde, kinesin-based fast axonal transport (FAT) by activating axonal protein phosphatase 1 (PP1) and glycogen synthase kinase 3 (GSK3), independent of microtubule binding. Defects in Tau are thought to be the cause of a number of neurodegenerative diseases, including frontotemporal dementia (FTD), pallido-ponto-nigral degeneration (PPND), Pick disease of the brain (PIDB), corticobasal degeneration (CBD), supranuclear palsy type 1 (PSNP1), Alzheimer disease, and Parkinson disease.

Immunogen

KLH-conjugated linear peptide corresponding to human Tau.

Application

Anti-Tau, PAD, clone TNT-1 detects levels of Tau proteins & has been published & validated for use in WB, IHC, IF & ELISA.
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases
Western Blot Analysis: 1 µg/mL from a representative lot detected Tau, PAD in 10 µg of human and mouse cortex tissue lysates.

Immunohistochemistry Analysis: A 1:2,000 dilution from a representative lot detected Tau, PAD in human Alzheimer′s brain tissue.

Immunohistochemistry Analysis: A representative lot from an independent laboratory detected Tau, PAD in normal and Alzheimer′s tissues from entorhinal cortex, hippocampus, inferior temporal gyrus, and superior gyrus of control (Kanaan, N. M., et al. (2011). J Neurosci. 31(27):9858-9868.).

ELISA Analysis: A representative lot from an independent laboratory detected Tau, PAD in recombinant Tau protein (Kanaan, N. M., et al. (2011). J Neurosci. 31(27):9858-9868.).

Quality

Evaluated by Western Blot in rat cortex tissue lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected Tau, PAD in 10 µg of rat cortex tissue lysate.

Target description

~55 kDa observed. Many isoforms are known to exist between 35-80 kDa An uncharacterized band may be observed in some cell lysates.

Linkage

Replaces: MAB10417

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
rat cortex tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuki Fujita et al.
STAR protocols, 1(3), 100111-100111 (2020-12-31)
Microglia, the resident immune cells of the central nervous system, accumulate along axons and support neuronal survival during the early postnatal period. Although methods have been developed to isolate microglia or neurons, the procedures for co-culture of these cells can
Hayk Davtyan et al.
Vaccine, 35(16), 2015-2024 (2017-03-23)
By the time clinical symptoms of Alzheimer's disease (AD) manifest in patients there is already substantial tau pathology in the brain. Recent evidence also suggests that tau pathology can become self-propagating, further accelerating disease progression. Over the last decade several
Michael G Agadjanyan et al.
Molecular neurodegeneration, 12(1), 33-33 (2017-05-06)
The experience from clinical trials indicates that anti-Aβ immunotherapy could be effective in early/pre-clinical stages of AD, whereas at the late stages promoting the clearing of Aβ alone may be insufficient to halt the disease progression. At the same time

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service