Skip to Content
Merck
All Photos(1)

Key Documents

MABF2042

Sigma-Aldrich

Anti-TSLP Receptor Antibody, clone 1A6

clone 1A6, from mouse

Synonym(s):

Cytokine receptor-like factor 2, Cytokine receptor-like 2, IL-XR, Thymic stromal lymphopoietin protein receptor, TSLPR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1A6, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

flow cytometry: suitable

isotype

IgG2aλ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... CRLF2(64109)

Related Categories

General description

Cytokine receptor-like factor 2 (UniProt: Q9HC73; also known as Cytokine receptor-like 2, IL-XR, Thymic stromal lymphopoietin protein receptor, TSLP receptor) is encoded by the CRLF2 (also known as CRL2, ILXR, TSLPR) gene (Gene ID: 64109) in human. TSLP receptor is a single-pass type I membrane protein that serves as a receptor for thymic stromal lymphopoietin. It forms a functional complex with TSLP and IL7R to stimulate cell proliferation via activation of STAT3 and STAT5. It is primarily expressed in dendrites and monocytes and its expression has also been reported in heart, skeletal muscle, kidney and liver. Its levels are up-regulated in activated peripheral monocytes and THP-1 cells. TSLP receptor is synthesized with a signal peptide (aa 1-21), which is subsequently cleaved off in the mature form. It contains an extracellular domain (aa 23-231), a transmembrane domain (aa 232-252), and a cytoplasmic domain (aa 253-371). It also has a WSXWS motif (aa 200-204) that is essential for proper protein folding and efficient intracellular transport and cell-surface binding. In addition, its Box 1 motif (aa 261-269) is reported to be important for its association with JAKs. Three isoforms of TSLP receptors have been described that are produced by alternative splicing.

Specificity

Clone 1A6 specifically detects human TSLP receptor protein. It targets an epitope with in the extracellular domain.

Immunogen

Epitope: extracellular domain
Fc-fusion protein of ectodomain region of human TSLP receptor protein.

Application

Anti-TSLP Receptor, clone 1A6, Cat. No. MABF2042, is a mouse monoclonal antibody that detects TSLP receptor and has been tested for use in Flow Cytometry.
Research Category
Inflammation & Immunology

Quality

Evaluated by Flow Cytometry in one million THP-1 monocytic peripheral blood cells.

Flow Cytometry Analysis: 1 µg of this antibody detected TSLP Receptor in one million THP-1 monocytic peripheral blood cells.

Target description

42.01 kDa calculated.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service